Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00966665_g1
          See other EIF2S2 GE Assays ›
          Gene Symbol:
          EIF2S2
          Gene Name:
          eukaryotic translation initiation factor 2 subunit beta
          Gene Aliases:
          EIF2, EIF2B, EIF2beta, PPP1R67, eIF-2-beta
          Chromosome Location:
          Chr.20: 34088298 - 34112356 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00966665_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EIF2S2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001316364.1 NP_001303293.1 2 - 3 440 150
          NM_003908.4 NP_003899.2 2 - 3 440 150
          XM_017028118.1 XP_016883607.1 2 - 3 602 150
          GenBank mRNA AB451237.1 - 1 - 2 194 150
          AB451362.1 - 1 - 2 194 150
          AK313025.1 - 2 - 3 311 150
          BC000461.2 - 2 - 3 330 2822324 150
          BC000934.2 - 2 - 3 305 3447202 150
          BC007888.2 - 1 - 2 121 4120898 150
          BT009795.1 - 1 - 2 194 150
          BX418731.2 - 2 - 3 310 150
          CR456734.1 - 1 - 2 194 150
          M29536.1 - 2 - 3 310 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008909.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000779.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC007888.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EIF2S2

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          male germ cell proliferation
          translational initiation
          male gonad development
          transmembrane transport
          RNA binding
          translation initiation factor activity
          protein binding
          translation factor activity, RNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012670.hg.1 ENST00000604970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011431.hg.1 ENST00000428356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011431.hg.1 ENST00000428356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013409.hg.1 ENST00000447161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010404.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010404.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010404.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010405.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010405.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010405.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010406.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010406.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010406.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010410.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010410.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010410.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009498.hg.1 ENST00000457635.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010407.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010407.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010407.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20010408.hg.1 TC20000779.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20010408.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20010408.hg.1 NM_003908.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009932.hg.1 ENST00000461070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009932.hg.1 ENST00000461070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009932.hg.1 ENST00000461070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009932.hg.1 ENST00000461070.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008618.hg.1 ENST00000527936.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162097.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162097.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162097.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162097.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162097.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC1000011431.hg.1 ENST00000428356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011431.hg.1 ENST00000428356.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162094.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162094.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162094.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162094.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162094.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162096.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162096.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162096.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162096.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162096.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162103.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162103.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162103.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162103.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162103.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009498.hg.1 ENST00000457635.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012670.hg.1 ENST00000604970.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162102.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162102.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162102.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162102.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162102.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC2000008909.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013409.hg.1 ENST00000447161.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162100.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162100.hg.1 NM_003908.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000162100.hg.1 TC2000008909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000162100.hg.1 ENST00000374980.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000162100.hg.1 NM_003908.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 161988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0