Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00969578_g1
          See other CNOT2 GE Assays ›
          Gene Symbol:
          CNOT2
          Gene Name:
          CCR4-NOT transcription complex subunit 2
          Gene Aliases:
          CDC36, HSPC131, NOT2, NOT2H
          Chromosome Location:
          Chr.12: 70242993 - 70354993 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00969578_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CNOT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199302.1 NP_001186231.1 13 - 14 1684 91
          NM_001199303.1 NP_001186232.1 12 - 13 1937 91
          NM_014515.5 NP_055330.1 12 - 13 1815 91
          NR_037615.1 13 - 14 2161 91
          XM_006719429.3 XP_006719492.1 13 - 14 2040 91
          XM_006719430.3 XP_006719493.1 13 - 14 1874 91
          XM_006719432.3 XP_006719495.1 11 - 12 1633 91
          XM_006719433.3 XP_006719496.1 9 - 10 1405 91
          XM_011538399.1 XP_011536701.1 12 - 13 1770 91
          XM_011538400.2 XP_011536702.1 13 - 14 2003 91
          XM_017019348.1 XP_016874837.1 14 - 15 2061 91
          XM_017019349.1 XP_016874838.1 14 - 15 2227 91
          XM_017019350.1 XP_016874839.1 14 - 15 1995 91
          XM_017019351.1 XP_016874840.1 14 - 15 2161 91
          XM_017019352.1 XP_016874841.1 15 - 16 2182 91
          XM_017019353.1 XP_016874842.1 10 - 11 1463 91
          XM_017019354.1 XP_016874843.1 10 - 11 1629 91
          GenBank mRNA AF113226.1 - 11 - 12 1342 91
          AF161480.1 - 12 - 13 1353 91
          AF180473.1 - 12 - 13 1348 91
          AK000662.1 - 12 - 13 1228 91
          AK095789.1 - 12 - 13 1900 91
          AK307479.1 - 13 - 14 2161 91
          AK315715.1 - 13 - 14 1685 91
          AL137674.1 - 4 - 5 364 91
          BC002597.2 - 12 - 13 1359 3143289 91
          BC011826.2 - 12 - 13 1367 3832035 91
          BX641116.1 - 3 - 4 3739 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008162.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000620.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CNOT2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          nuclear-transcribed mRNA poly(A) tail shortening
          trophectodermal cell differentiation
          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          positive regulation of cytoplasmic mRNA processing body assembly
          negative regulation of translation
          gene silencing by RNA
          negative regulation of intracellular estrogen receptor signaling pathway
          RNA phosphodiester bond hydrolysis, exonucleolytic
          regulation of stem cell population maintenance
          transcription corepressor binding
          transcription coregulator activity
          poly(A)-specific ribonuclease activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 ENST00000229195.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 ENST00000418359.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 ENST00000551483.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 NM_001199302.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 NM_001199303.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 NM_014515.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 NR_037615.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12008248.hg.1 TC12000620.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000229195.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000418359.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000546673.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548159.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548338.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548939.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000549709.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551043.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551434.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551483.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551710.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000552599.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_001199302.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_001199303.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_014515.5 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NR_037615.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 TC1200008162.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000546673.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000229195.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000418359.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548159.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548338.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000548939.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000549709.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551043.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551434.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551483.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000551710.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 ENST00000552599.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_001199302.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_001199303.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NM_014515.5 Refseq
          Affymetrix Human Clariom D Assay PSR1200159947.hg.1 NR_037615.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline