Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00969920_s1
          See other APOOP5 GE Assays ›
          Gene Symbol:
          APOOP5
          Gene Name:
          apolipoprotein O pseudogene 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 59754132 - 59755191 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00969920_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: APOOP5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028471.1 1 141
          GenBank mRNA AI016649.1 - 1 91 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010509.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001160.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3220670 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40950 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3220670 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16901870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588740 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588740 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16901870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10048392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30588729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16016145.hg.1 NR_028471.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16016145.hg.1 TC16001160.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1600180850.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180850.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180850.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180850.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180850.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180851.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564533.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010509.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180854.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180854.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180854.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 ENST00000565921.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180852.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180849.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180849.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180849.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180853.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180853.hg.1 NR_028471.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600180853.hg.1 TC1600010509.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600180853.hg.1 ENST00000564761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600180853.hg.1 NR_028471.1 Refseq
          Human Genome U95 Set 71743_at - -
          Human Genome U95 Set 71743_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053807 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053807 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0