Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00971622_m1
          See other LIMCH1 GE Assays ›
          Gene Symbol:
          LIMCH1
          Gene Name:
          LIM and calponin homology domains 1
          Gene Aliases:
          LIMCH1A, LMO7B
          Chromosome Location:
          Chr.4: 41359607 - 41700044 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00971622_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LIMCH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001112718.2 NP_001106189.1 17 - 18 2341 131
          NM_001112719.2 NP_001106190.1 12 - 13 2126 131
          NM_001112720.2 NP_001106191.1 11 - 12 2090 131
          NM_001289122.1 NP_001276051.1 17 - 18 2380 206
          NM_001289124.1 NP_001276053.1 17 - 18 2425 206
          XM_005248058.1 XP_005248115.1 24 - 25 3973 131
          XM_005248060.1 XP_005248117.1 24 - 25 3973 131
          XM_005248067.2 XP_005248124.1 22 - 23 4847 131
          XM_005248072.1 XP_005248129.1 17 - 18 2341 131
          XM_005248074.1 XP_005248131.1 16 - 17 2305 131
          XM_005248075.1 XP_005248132.1 16 - 17 2305 131
          XM_006713996.1 XP_006714059.1 24 - 25 3973 131
          XM_011513643.1 XP_011511945.1 25 - 26 4048 206
          XM_011513656.1 XP_011511958.1 12 - 13 2000 131
          XM_011513657.1 XP_011511959.1 11 - 12 1964 131
          XM_011513658.2 XP_011511960.1 16 - 17 2566 131
          XM_017007894.1 XP_016863383.1 25 - 26 4048 206
          XM_017007899.1 XP_016863388.1 17 - 18 2341 131
          XM_017007904.1 XP_016863393.1 12 - 13 2000 131
          GenBank mRNA AK027231.1 - 4 - 5 346 131
          AK294774.1 - 11 - 12 131
          AK295784.1 - 10 - 11 1679 131
          AK295836.1 - 11 - 12 1789 131
          AK298915.1 - 12 - 13 131
          AK299297.1 - 11 - 12 131
          AK299835.1 - 11 - 12 1824 131
          AK302674.1 - 4 - 5 131
          AK309846.1 - 11 - 12 2032 131
          AL831962.2 - 22 - 23 131
          BC023546.2 - 14 - 15 4110496 131
          BC029735.1 - 2 - 3 96 131
          BX537916.1 - 11 - 12 131
          BX640692.1 - 10 - 11 131
          CR749205.1 - 12 - 13 131
          CR936601.1 - 17 - 18 2351 206
          CR936610.1 - 17 - 18 2426 206
          CR936661.1 - 16 - 17 131
          CR936664.1 - 17 - 18 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007345.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000254.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          actomyosin structure organization
          actin binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000313860.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000313875.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000381753.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000396595.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000503057.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000508501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000509277.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000511496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000512820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000512946.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000513024.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000514096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 ENST00000537405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 NM_014988.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003727.hg.1 TC04000254.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000313860.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000381753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000396595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000503057.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000508466.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000508501.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000509277.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000511496.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000512820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000512946.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000513024.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000514096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001289122.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001289124.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_014988.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 TC0400007345.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000503057.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000313860.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000381753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000396595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000508466.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000508501.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000509277.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000511496.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000512820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000512946.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000513024.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 ENST00000514096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001289122.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_001289124.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400150247.hg.1 NM_014988.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0