Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00971626_m1
          See other LIMCH1 GE Assays ›
          Gene Symbol:
          LIMCH1
          Gene Name:
          LIM and calponin homology domains 1
          Gene Aliases:
          LIMCH1A, LMO7B
          Chromosome Location:
          Chr.4: 41359607 - 41700044 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00971626_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LIMCH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001112717.2 NP_001106188.1 23 - 24 2993 67
          NM_001112718.2 NP_001106189.1 23 - 24 2990 67
          NM_001112719.2 NP_001106190.1 18 - 19 2775 67
          NM_001112720.2 NP_001106191.1 17 - 18 2739 67
          NM_001289122.1 NP_001276051.1 23 - 24 3029 67
          NM_001289124.1 NP_001276053.1 23 - 24 3074 67
          XM_005248075.1 XP_005248132.1 22 - 23 2954 67
          XM_006713996.1 XP_006714059.1 30 - 31 4622 67
          XM_011513646.1 XP_011511948.1 31 - 32 4700 67
          XM_011513658.2 XP_011511960.1 22 - 23 3215 67
          XM_017007896.1 XP_016863385.1 30 - 31 4616 67
          XM_017007897.1 XP_016863386.1 29 - 30 4541 67
          XM_017007902.1 XP_016863391.1 22 - 23 2909 67
          XM_017007905.1 XP_016863394.1 18 - 19 2652 67
          GenBank mRNA AK027231.1 - 10 - 11 995 67
          AK299297.1 - 17 - 18 2609 67
          AK302674.1 - 10 - 11 1241 67
          AK309846.1 - 17 - 18 2681 67
          BC023546.2 - 20 - 21 2843 4110496 67
          BC029735.1 - 8 - 9 745 67
          BC095394.1 - 13 - 14 1290 67
          BX537916.1 - 17 - 18 2609 67
          BX640692.1 - 16 - 17 2367 67
          CR749205.1 - 18 - 19 2645 67
          CR933645.1 - 23 - 24 2994 67
          CR936601.1 - 23 - 24 3000 67
          CR936610.1 - 23 - 24 3075 67
          CR936658.1 - 20 - 21 2741 67
          CR936661.1 - 22 - 23 2894 67
          CR936664.1 - 23 - 24 2991 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007345.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000254.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          actomyosin structure organization
          actin binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000313860.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000313875.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000381753.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000396595.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000503057.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000508501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000509277.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000511496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000512632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000512820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000512946.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000513024.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000514096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 ENST00000537405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 NM_014988.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04003734.hg.1 TC04000254.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000313860.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000381753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000396595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000503057.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000508466.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000508501.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000509277.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000511496.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512632.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512946.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000513024.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000514096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001289122.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001289124.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_014988.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 TC0400007345.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000313860.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000381753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000396595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000503057.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000508466.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000508501.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000509277.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000511496.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512632.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000512946.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000513024.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 ENST00000514096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112717.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112718.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112719.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001112720.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001289122.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_001289124.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400150254.hg.1 NM_014988.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0