Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00972074_m1
See other MYEF2 GE Assays ›
Gene Symbol:
MYEF2
Gene Name:
myelin expression factor 2
Gene Aliases:
HsT18564, MEF-2, MST156, MSTP156, myEF-2
Chromosome Location:
Chr.15: 48134632 - 48178517 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00972074_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MYEF2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001301210.1
NP_001288139.1
6 - 7
841
71
NM_016132.4
NP_057216.2
6 - 7
841
71
NR_125408.1
5 - 6
788
71
XM_005254422.4
XP_005254479.2
6 - 7
997
71
XM_005254424.4
XP_005254481.2
6 - 7
811
71
XM_005254425.4
XP_005254482.2
6 - 7
997
71
XM_005254427.4
XP_005254484.1
6 - 7
997
71
XM_006720553.3
XP_006720616.1
5 - 6
789
71
XM_011521657.2
XP_011519959.1
2 - 3
339
71
XM_017022285.1
XP_016877774.1
6 - 7
997
71
XM_017022286.1
XP_016877775.1
6 - 7
997
71
XM_017022287.1
XP_016877776.1
6 - 7
997
71
XM_017022288.1
XP_016877777.1
5 - 6
789
71
XM_017022289.1
XP_016877778.1
5 - 6
789
71
XM_017022290.1
XP_016877779.1
5 - 6
789
71
XM_017022291.1
XP_016877780.1
5 - 6
789
71
XM_017022292.1
XP_016877781.1
5 - 6
789
71
GenBank mRNA
AB037762.1
-
6 - 7
779
71
AF190159.1
-
6 - 7
603
71
AK023133.1
-
6 - 7
833
71
BC068523.1
-
5 - 6
743
71
BC152420.1
-
6 - 7
779
8862532
71
BC152451.1
-
6 - 7
779
8862583
71
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene MYEF2
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
myotube differentiation
neuron differentiation
nucleotide binding
DNA binding
poly(A) RNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0