Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00972295_m1
          See other MKL2 GE Assays ›
          Gene Symbol:
          MKL2
          Gene Name:
          MKL1/myocardin like 2
          Gene Aliases:
          MRTF-B, MRTFB, NPD001
          Chromosome Location:
          Chr.16: 14071076 - 14266779 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00972295_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MKL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001308142.1 NP_001295071.1 15 - 16 2885 79
          NM_014048.4 NP_054767.3 15 - 16 2736 79
          XM_005255452.3 XP_005255509.1 16 - 17 3040 79
          XM_005255453.4 XP_005255510.1 15 - 16 3524 79
          XM_005255455.3 XP_005255512.1 13 - 14 2870 79
          XM_006720908.3 XP_006720971.1 16 - 17 3686 79
          XM_006720909.3 XP_006720972.1 16 - 17 3015 79
          XM_006720912.3 XP_006720975.1 13 - 14 4825 79
          XM_006720914.2 XP_006720977.1 16 - 17 2890 79
          XM_011522568.2 XP_011520870.1 15 - 16 2855 79
          XM_011522569.2 XP_011520871.1 13 - 14 2652 79
          XM_017023501.1 XP_016878990.1 15 - 16 2851 79
          XM_017023502.1 XP_016878991.1 15 - 16 2908 79
          XM_017023503.1 XP_016878992.1 14 - 15 6053 79
          XM_017023504.1 XP_016878993.1 13 - 14 2720 79
          XM_017023505.1 XP_016878994.1 13 - 14 4675 79
          GenBank mRNA AB033069.2 - 7 - 8 1892 79
          AK127530.1 - 4 - 5 1161 79
          AK294765.1 - 15 - 16 2877 79
          AY374297.1 - 15 - 16 2713 79
          BC136260.1 - 13 - 14 2730 9020247 79
          BC171750.1 - 13 - 14 2730 9053251 79
          BX641041.1 - 3 - 4 1973 79
          CR749797.1 - 4 - 5 1431 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC16000171.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MKL2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          muscle organ development
          positive regulation of striated muscle tissue development
          positive regulation of transcription by RNA polymerase II
          smooth muscle cell differentiation
          cell-cell adhesion
          transcription coactivator activity
          protein binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16002623.hg.1 ENST00000318282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002623.hg.1 ENST00000341243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002623.hg.1 NM_014048.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16002623.hg.1 TC16000171.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix