Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00974739_m1
See other UNC79 GE Assays ›
Gene Symbol:
UNC79
Gene Name:
unc-79 homolog (C. elegans)
Gene Aliases:
KIAA1409
Chromosome Location:
Chr.14: 93333219 - 93710473 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00974739_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: UNC79
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_011537018.2
XP_011535320.1
30 - 31
6986
112
XM_011537020.2
XP_011535322.1
29 - 30
6920
112
XM_011537021.2
XP_011535323.1
30 - 31
4537
112
XM_011537022.2
XP_011535324.1
30 - 31
6986
112
XM_011537023.2
XP_011535325.1
30 - 31
6827
112
XM_011537024.2
XP_011535326.1
29 - 30
6764
112
XM_011537025.2
XP_011535327.1
30 - 31
6986
112
XM_011537026.2
XP_011535328.1
30 - 31
7479
112
XM_011537027.2
XP_011535329.1
30 - 31
4504
112
XM_011537028.2
XP_011535330.1
29 - 30
4503
112
XM_011537029.2
XP_011535331.1
21 - 22
3452
112
XM_017021506.1
XP_016876995.1
30 - 31
6884
112
XM_017021507.1
XP_016876996.1
30 - 31
6986
112
XM_017021509.1
XP_016876998.1
27 - 28
6827
112
XM_017021510.1
XP_016876999.1
29 - 30
6920
112
XM_017021511.1
XP_016877000.1
25 - 26
3780
112
XM_017021512.1
XP_016877001.1
29 - 30
4471
112
XM_017021515.1
XP_016877004.1
30 - 31
4385
112
XM_017021518.1
XP_016877007.1
24 - 25
4908
112
XM_017021519.1
XP_016877008.1
20 - 21
4002
112
XM_017021520.1
XP_016877009.1
14 - 15
2307
112
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Homo sapiens
Probe spans exons
Gene Ontology Categories:
Function(s)
ion transmembrane transport
multicellular organism growth
behavioral response to ethanol
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0