Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00974761_m1
          See other UNC79 GE Assays ›
          Gene Symbol:
          UNC79
          Gene Name:
          unc-79 homolog (C. elegans)
          Gene Aliases:
          KIAA1409
          Chromosome Location:
          Chr.14: 93333219 - 93710473 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00974761_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UNC79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020818.3 NP_065869.3 48 - 49 7796 140
          XM_011537018.2 XP_011535320.1 50 - 51 10429 140
          XM_011537020.2 XP_011535322.1 49 - 50 10363 140
          XM_011537021.2 XP_011535323.1 50 - 51 7980 140
          XM_011537022.2 XP_011535324.1 50 - 51 10273 140
          XM_011537023.2 XP_011535325.1 50 - 51 10270 140
          XM_011537024.2 XP_011535326.1 49 - 50 10207 140
          XM_011537026.2 XP_011535328.1 50 - 51 10922 140
          XM_011537027.2 XP_011535329.1 50 - 51 7947 140
          XM_011537028.2 XP_011535330.1 49 - 50 7946 140
          XM_011537029.2 XP_011535331.1 41 - 42 6895 140
          XM_017021506.1 XP_016876995.1 50 - 51 10327 140
          XM_017021507.1 XP_016876996.1 49 - 50 10312 140
          XM_017021508.1 XP_016876997.1 49 - 50 10279 140
          XM_017021509.1 XP_016876998.1 47 - 48 10270 140
          XM_017021510.1 XP_016876999.1 48 - 49 10246 140
          XM_017021511.1 XP_016877000.1 45 - 46 7223 140
          XM_017021512.1 XP_016877001.1 49 - 50 7914 140
          XM_017021513.1 XP_016877002.1 48 - 49 10213 140
          XM_017021514.1 XP_016877003.1 48 - 49 10162 140
          XM_017021515.1 XP_016877004.1 50 - 51 7828 140
          XM_017021516.1 XP_016877005.1 47 - 48 10096 140
          XM_017021517.1 XP_016877006.1 47 - 48 9937 140
          XM_017021518.1 XP_016877007.1 44 - 45 8351 140
          XM_017021519.1 XP_016877008.1 40 - 41 7445 140
          XM_017021520.1 XP_016877009.1 34 - 35 5750 140
          GenBank mRNA AB037830.2 - 46 - 47 7150 140
          AK090721.1 - 11 - 12 1205 140
          AK126626.1 - 2 - 3 3041 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008034.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000577.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UNC79

          Gene Ontology Categories:

          Function(s)

          ion transmembrane transport
          multicellular organism growth
          behavioral response to ethanol

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 ENST00000256339.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 ENST00000393151.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 ENST00000393153.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 ENST00000553484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 ENST00000555664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 NM_020818.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14007111.hg.1 TC14000577.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000256339.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000393151.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000553484.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000554549.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000555664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000621021.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 NM_020818.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 TC1400008034.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000256339.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000393151.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000553484.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000554549.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000555664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 ENST00000621021.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400156960.hg.1 NM_020818.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0