Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00974917_m1
          See other SHANK2 GE Assays ›
          Gene Symbol:
          SHANK2
          Gene Name:
          SH3 and multiple ankyrin repeat domains 2
          Gene Aliases:
          AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
          Chromosome Location:
          Chr.11: 70467856 - 71252724 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00974917_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SHANK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012309.4 NP_036441.2 23 - 24 2678 71
          NM_133266.4 NP_573573.2 9 - 10 875 71
          XM_005277932.3 XP_005277989.1 14 - 15 1683 71
          XM_017017387.1 XP_016872876.1 24 - 25 2783 71
          XM_017017388.1 XP_016872877.1 24 - 25 2805 71
          XM_017017390.1 XP_016872879.1 11 - 12 3916 71
          GenBank mRNA AB028945.1 - 6 - 7 412 71
          AB208025.1 - 23 - 24 2644 71
          AB208026.1 - 14 - 15 1466 71
          AB208027.1 - 9 - 10 853 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100011485.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SHANK2

          Gene Ontology Categories:

          Function(s) Process(es)

          synapse assembly
          learning
          adult behavior
          social behavior
          long-term synaptic potentiation
          long-term synaptic depression
          vocalization behavior
          protein binding
          SH3 domain binding
          synaptic receptor adaptor activity
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000338508.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000409161.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000423696.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000424924.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000449833.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000601538.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 NM_012309.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 NM_133266.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 TC1100011485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000338508.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000409161.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000423696.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000424924.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000449833.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 ENST00000601538.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 NM_012309.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100188425.hg.1 NM_133266.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline