Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00974994_s1
          See other LOC338620 GE Assays ›
          Gene Symbol:
          LOC338620
          Gene Name:
          uncharacterized LOC338620
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00974994_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC338620
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC043009.1 - 1 964 117
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA426512.1 - 1 Chr. 10 - 32887255 - 32887927 [+] on Build GRCh38 117
          GenBankmRNA AI627380.1 - 1 Chr. 10 - 32887255 - 32887625 [+] on Build GRCh38 117
          GenBankmRNA BU632601.1 - 1 Chr. 10 - 32887255 - 32887911 [+] on Build GRCh38 117
          GenBankmRNA BF432102.1 - 1 Chr. 10 - 32887255 - 32887722 [+] on Build GRCh38 117
          GenBankmRNA BU633613.1 - 1 Chr. 10 - 32887255 - 32887967 [+] on Build GRCh38 117
          GenBankmRNA AA406479.1 - 1 Chr. 10 - 32887262 - 32887556 [+] on Build GRCh38 117
          GenBankmRNA AA236788.1 - 1 Chr. 10 - 32887262 - 32887747 [+] on Build GRCh38 117
          GenBankmRNA HY212242.1 - 1 Chr. 10 - 32887263 - 32887699 [+] on Build GRCh38 117
          GenBankmRNA CA426358.1 - 1 Chr. 10 - 32887255 - 32887964 [+] on Build GRCh38 117
          GenBankmRNA AW300483.1 - 1 Chr. 10 - 32887262 - 32887700 [+] on Build GRCh38 117
          GenBankmRNA AI435839.1 - 1 Chr. 10 - 32887262 - 32887807 [+] on Build GRCh38 117
          GenBankmRNA AI588976.1 - 1 Chr. 10 - 32887257 - 32887746 [+] on Build GRCh38 117
          GenBankmRNA CA422751.1 - 1 Chr. 10 - 32887255 - 32887910 [+] on Build GRCh38 117

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coiled-coil domain containing 7 CCDC7 BIOT2,
          BioT2-A,
          BioT2-B,
          BioT2-C,
          C10orf68
          -4.4kb Sense Chr. 10 - 32446082 - 32882864 on NCBI Build GRCh38
          Downstream integrin subunit beta 1 ITGB1 CD29,
          FNRB,
          GPIIA,
          MDF2,
          MSK12,
          VLA-BETA,
          VLAB
          12.4kb Anti-sense Chr. 10 - 32900318 - 32958365 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3799692 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P97780 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21479 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53601 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3799692 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P97780 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P97780 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 230930_PM_at - -
          Human Genome U133 Plus 2.0 Array 230930_PM_at - -
          Human Genome U133 Plus 2.0 Array 230930_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230930_at9 - -
          Human Genome U95 Set 55363_at - -
          Human Genome U95 Set 55363_PM_at - -
          Human Genome U133 Set 230930_at - -
          Human Genome U133 Set 230930_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 157049 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1861057 BC043009.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1861057 BC043009.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1861057 BC043009.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6100630 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_113091 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1861057 BC043009.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1861057 BC043009.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1861057 BC043009.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0