Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014648.3 | NP_055463.1 | 18 - 19 | 2450 | 136 | |
XM_005247914.3 | XP_005247971.1 | 18 - 19 | 2249 | 136 | ||
XM_005247915.3 | XP_005247972.1 | 18 - 19 | 2219 | 136 | ||
XM_005247916.3 | XP_005247973.1 | 19 - 20 | 2391 | 136 | ||
XM_005247917.3 | XP_005247974.1 | 18 - 19 | 2180 | 136 | ||
XM_005247918.3 | XP_005247975.1 | 18 - 19 | 2667 | 136 | ||
XM_005247919.3 | XP_005247976.1 | 17 - 18 | 1801 | 136 | ||
XM_017007540.1 | XP_016863029.1 | 18 - 19 | 2296 | 136 | ||
GenBank mRNA | AB014575.1 | - | 18 - 19 | 2157 | 136 | |
AF279370.1 | - | 3 - 4 | 109 | 136 | ||
AK023138.1 | - | 18 - 19 | 2218 | 136 | ||
AK294967.1 | - | 7 - 8 | 513 | 136 | ||
AK303942.1 | - | 5 - 6 | 653 | 136 | ||
AY227652.1 | - | 5 - 6 | 361 | 136 | ||
AY227653.1 | - | 9 - 10 | 1213 | 136 | ||
BC063882.1 | - | 18 - 19 | 2203 | 30387514 | 136 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |