Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00980084_m1
          See other ERN1 GE Assays ›
          Gene Symbol:
          ERN1
          Gene Name:
          endoplasmic reticulum to nucleus signaling 1
          Gene Aliases:
          IRE1, IRE1P, IRE1a, hIRE1p
          Chromosome Location:
          Chr.17: 64039142 - 64130819 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00980084_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001433.3 NP_001424.3 13 - 14 1785 143
          XM_017024347.1 XP_016879836.1 13 - 14 3139 143
          XM_017024348.1 XP_016879837.1 12 - 13 3037 143
          GenBank mRNA AB209869.1 - 13 - 14 1768 143
          AF059198.1 - 13 - 14 1768 143
          AK292403.1 - 13 - 14 1811 143
          BC130405.1 - 13 - 14 1718 40146436 143
          BC130407.1 - 13 - 14 1718 40146438 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011436.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001796.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ERN1

          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA cleavage
          mRNA catabolic process
          protein phosphorylation
          activation of signaling protein activity involved in unfolded protein response
          cell cycle arrest
          activation of JUN kinase activity
          regulation of macroautophagy
          positive regulation of RNA splicing
          response to endoplasmic reticulum stress
          cellular response to vascular endothelial growth factor stimulus
          peptidyl-serine autophosphorylation
          IRE1-mediated unfolded protein response
          protein autophosphorylation
          mRNA splicing, via endonucleolytic cleavage and ligation
          mRNA endonucleolytic cleavage involved in unfolded protein response
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to glucose stimulus
          RNA phosphodiester bond hydrolysis, endonucleolytic
          positive regulation of endoplasmic reticulum unfolded protein response
          insulin metabolic process
          peptidyl-serine trans-autophosphorylation
          magnesium ion binding
          endoribonuclease activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          enzyme binding
          Hsp70 protein binding
          identical protein binding
          protein homodimerization activity
          ADP binding
          Hsp90 protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0