Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00981458_m1
          See other MYLK GE Assays ›
          Gene Symbol:
          MYLK
          Gene Name:
          myosin light chain kinase
          Gene Aliases:
          AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
          Chromosome Location:
          Chr.3: 123612296 - 123884302 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00981458_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYLK
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321309.1 NP_001308238.1 25 - 26 4493 90
          NM_053025.3 NP_444253.3 26 - 27 4701 90
          NM_053026.3 NP_444254.3 25 - 26 4494 90
          NM_053027.3 NP_444255.3 26 - 27 4701 90
          NM_053028.3 NP_444256.3 25 - 26 4494 90
          XM_011512860.2 XP_011511162.1 23 - 24 4422 90
          XM_017006469.1 XP_016861958.1 9 - 10 1980 90
          XM_017006470.1 XP_016861959.1 8 - 9 968 90
          XM_017006471.1 XP_016861960.1 8 - 9 1018 90
          GenBank mRNA AB037663.1 - 9 - 10 1777 90
          AF069601.2 - 23 - 24 4331 90
          AF069602.1 - 24 - 25 4538 90
          AF069603.1 - 23 - 24 4331 90
          AK300610.1 - 8 - 9 951 90
          AY339601.1 - 23 - 24 4419 90
          AY424269.1 - 22 - 23 4214 90
          AY424270.1 - 23 - 24 4421 90
          U48959.2 - 24 - 25 4538 90
          X85337.1 - 10 - 11 2034 90
          X90870.1 - 1 - 2 54 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300012212.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001713.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYLK

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          protein phosphorylation
          muscle contraction
          smooth muscle contraction
          tonic smooth muscle contraction
          positive regulation of cell migration
          bleb assembly
          positive regulation of calcium ion transport
          aorta smooth muscle tissue morphogenesis
          cellular hypotonic response
          positive regulation of wound healing
          positive regulation of mitophagy in response to mitochondrial depolarization
          xenophagy
          actin binding
          myosin light chain kinase activity
          protein binding
          calmodulin binding
          ATP binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000346322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000354792.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000359169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000360304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000360772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 ENST00000475616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 NM_053025.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 NM_053026.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 NM_053027.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 NM_053028.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03031055.hg.1 TC03001713.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000346322.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000354792.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000359169.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000360304.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000360772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000464489.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000475616.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000513111.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053025.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053026.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053027.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053028.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 TC0300012212.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000360304.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000346322.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000354792.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000359169.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000360772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000464489.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000475616.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 ENST00000513111.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053025.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053026.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053027.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300191069.hg.1 NM_053028.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline