Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00985367_m1
          See other ITGA2 GE Assays ›
          Gene Symbol:
          ITGA2
          Gene Name:
          integrin subunit alpha 2
          Gene Aliases:
          BR, CD49B, GPIa, HPA-5, VLA-2, VLAA2
          Chromosome Location:
          Chr.5: 52989326 - 53094779 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00985367_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002203.3 NP_002194.2 14 - 15 1950 92
          NR_073103.1 14 - 15 1950 92
          NR_073104.1 14 - 15 1950 92
          NR_073106.1 14 - 15 1950 92
          NR_073107.1 13 - 14 1829 92
          GenBank mRNA AK307952.1 - 14 - 15 1939 92
          BC143505.1 - 14 - 15 1836 92
          BC143508.1 - 14 - 15 1836 92
          BC143511.1 - 14 - 15 1836 92
          BC143512.1 - 13 - 14 1715 92
          X17033.1 - 14 - 15 1855 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500007368.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000218.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITGA2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of leukocyte migration
          substrate-dependent cell migration
          hypotonic response
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          female pregnancy
          blood coagulation
          cell population proliferation
          response to xenobiotic stimulus
          animal organ morphogenesis
          positive regulation of epithelial cell migration
          positive regulation of alkaline phosphatase activity
          response to amine
          response to muscle activity
          positive regulation of smooth muscle cell migration
          extracellular matrix organization
          mammary gland development
          positive regulation of cell projection organization
          cell-substrate adhesion
          positive regulation of collagen biosynthetic process
          positive regulation of collagen binding
          response to L-ascorbic acid
          cell adhesion mediated by integrin
          collagen-activated signaling pathway
          positive regulation of DNA binding
          skin morphogenesis
          establishment of protein localization
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of smooth muscle contraction
          viral entry into host cell
          focal adhesion assembly
          mesodermal cell differentiation
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of positive chemotaxis
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of transmission of nerve impulse
          positive regulation of phagocytosis, engulfment
          hepatocyte differentiation
          response to parathyroid hormone
          cellular response to mechanical stimulus
          cellular response to estradiol stimulus
          virus receptor activity
          integrin binding
          protein binding
          collagen binding
          collagen receptor activity
          laminin binding
          protein-containing complex binding
          metal ion binding
          protein heterodimerization activity
          collagen binding involved in cell-matrix adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05002690.hg.1 ENST00000296585.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002690.hg.1 NM_002203.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05002690.hg.1 TC05000218.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000296585.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000503810.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000509814.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000509960.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000510722.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000513685.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NM_002203.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073103.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073104.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073105.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073107.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 TC0500007368.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000509960.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000296585.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000503810.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000509814.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000510722.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 ENST00000513685.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NM_002203.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073103.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073104.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073105.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073106.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149800.hg.1 NR_073107.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0