Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00987262_g1
          See other PKM GE Assays ›
          Gene Symbol:
          PKM
          Gene Name:
          pyruvate kinase, muscle
          Gene Aliases:
          CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1
          Chromosome Location:
          Chr.15: 72199029 - 72231624 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00987262_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PKM
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001206797.2 NP_001193726.1 8 - 9 1603 95
          NM_001206798.2 NP_001193727.1 9 - 10 1850 95
          NM_002654.5 NP_002645.3 9 - 10 1825 95
          XM_005254443.1 XP_005254500.1 10 - 11 1549 95
          XM_005254445.4 XP_005254502.1 9 - 10 1386 95
          XM_006720570.1 XP_006720633.1 10 - 11 1646 95
          XM_011521670.2 XP_011519972.1 9 - 10 1611 95
          GenBank mRNA AF025439.1 - 2 - 3 202 95
          AK222927.1 - 9 - 10 1391 95
          AK297951.1 - 1169 95
          AK300800.1 - 9 - 10 1416 95
          AK312253.1 - 9 - 10 1391 95
          BC000481.2 - 9 - 10 1407 2964687 95
          BC007640.1 - 9 - 10 1391 3533876 95
          BC007952.2 - 9 - 10 1403 4299213 95
          BC012811.2 - 9 - 10 1403 2958817 95
          BC019265.2 - 4 - 5 739 95
          BC023328.1 - 5 - 6 775 95
          BC035198.1 - 9 - 10 1357 3859987 95
          EU794631.1 - 9 - 10 1544 95
          M23725.1 - 9 - 10 1412 95
          M26252.1 - 9 - 10 1392 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009952.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PKM

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          ATP biosynthetic process
          response to nutrient
          response to gravity
          programmed cell death
          response to muscle inactivity
          animal organ regeneration
          response to insulin
          pyruvate biosynthetic process
          skeletal muscle tissue regeneration
          canonical glycolysis
          cell-cell adhesion
          magnesium ion binding
          RNA binding
          pyruvate kinase activity
          protein binding
          ATP binding
          kinase activity
          MHC class II protein complex binding
          potassium ion binding
          ADP binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800008001.hg.1 ENST00000523134.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008001.hg.1 ENST00000523134.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000319622.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000335181.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000389093.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000449901.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000561609.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000564440.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565143.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565154.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565184.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000567118.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000568459.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000568883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206796.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206797.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206799.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_002654.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_182470.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_182471.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 TC1500009952.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000564440.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000319622.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000335181.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000389093.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000449901.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000561609.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565143.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565154.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000565184.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000567118.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000568459.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 ENST00000568883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206796.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206797.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_001206799.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_002654.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_182470.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500173886.hg.1 NM_182471.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0