Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105515.2 | NP_001098985.1 | 17 - 18 | 2343 | 86 | |
NM_001301829.1 | NP_001288758.1 | 16 - 17 | 2202 | 86 | ||
NM_001301830.1 | NP_001288759.1 | 16 - 17 | 2118 | 86 | ||
NM_005845.4 | NP_005836.2 | 17 - 18 | 2343 | 86 | ||
XM_005254025.2 | XP_005254082.1 | 18 - 19 | 2403 | 86 | ||
XM_017020319.1 | XP_016875808.1 | 18 - 19 | 2489 | 86 | ||
XM_017020320.1 | XP_016875809.1 | 17 - 18 | 2447 | 86 | ||
XM_017020321.1 | XP_016875810.1 | 7 - 8 | 1858 | 86 | ||
XM_017020322.1 | XP_016875811.1 | 18 - 19 | 2352 | 86 | ||
GenBank mRNA | AB208973.1 | - | 15 - 16 | 1962 | 86 | |
AF071202.1 | - | 17 - 18 | 2326 | 86 | ||
AF071203.1 | - | 9 - 10 | 981 | 86 | ||
AF541977.1 | - | 16 - 17 | 2075 | 86 | ||
AK290317.1 | - | 17 - 18 | 2343 | 86 | ||
AK296247.1 | - | 16 - 17 | 2105 | 86 | ||
AY081219.1 | - | 17 - 18 | 2232 | 86 | ||
AY207008.1 | - | 17 - 18 | 2216 | 86 | ||
BC041560.1 | - | 17 - 18 | 2313 | 5812052 | 86 | |
BX647118.1 | - | 7 - 8 | 718 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |