Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00989808_m1
          See other FRZB GE Assays ›
          Gene Symbol:
          FRZB
          Gene Name:
          frizzled-related protein
          Gene Aliases:
          FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
          Chromosome Location:
          Chr.2: 182833275 - 182866770 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00989808_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FRZB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001463.3 NP_001454.2 1 - 2 697 70
          GenBank mRNA AK130009.1 - 2 - 3 349 70
          AK312741.1 - 1 - 2 576 70
          BC027855.1 - 1 - 2 549 5221358 70
          BT019883.1 - 1 - 2 479 70
          U24163.1 - 1 - 2 687 70
          U68057.1 - 1 - 2 539 70
          U91903.1 - 1 - 2 548 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015136.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002591.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FRZB

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          G protein-coupled receptor signaling pathway
          negative regulation of cell population proliferation
          negative regulation of cell development
          neural crest cell differentiation
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          non-canonical Wnt signaling pathway
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          convergent extension involved in organogenesis
          negative regulation of cartilage development
          somite development
          negative regulation of hepatocyte differentiation
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          G protein-coupled receptor activity
          Wnt-protein binding
          Wnt-activated receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02041562.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041562.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041562.hg.1 TC02002591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02041564.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041564.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041564.hg.1 TC02002591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041563.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041563.hg.1 NM_001463.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041563.hg.1 TC02002591.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200210920.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210920.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210920.hg.1 TC0200015136.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200210920.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210920.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015136.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210919.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210919.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210919.hg.1 TC0200015136.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200210919.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210919.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210918.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210918.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210918.hg.1 TC0200015136.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200210918.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210918.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210921.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210921.hg.1 NM_001463.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200210921.hg.1 TC0200015136.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200210921.hg.1 ENST00000295113.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200210921.hg.1 NM_001463.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 149805 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0