Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00990551_s1
          See other EXOSC6 GE Assays ›
          Gene Symbol:
          EXOSC6
          Gene Name:
          exosome component 6
          Gene Aliases:
          EAP4, MTR3, Mtr3p, hMtr3p, p11
          Chromosome Location:
          Chr.16: 70250231 - 70251930 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00990551_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EXOSC6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_058219.2 NP_478126.1 1 951 65
          GenBank mRNA BC052252.1 - 1 951 6256140 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010751.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EXOSC6

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          rRNA catabolic process
          rRNA 3'-end processing
          nuclear-transcribed mRNA catabolic process, exonucleolytic, 3'-5'
          U4 snRNA 3'-end processing
          regulation of mRNA stability
          exonucleolytic catabolism of deadenylated mRNA
          DNA deamination
          isotype switching
          nuclear mRNA surveillance
          polyadenylation-dependent snoRNA 3'-end processing
          RNA binding
          exoribonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10095145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30636422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10095142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10095150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30636415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16909955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30636397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10095146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30636432 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 210563 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P39639 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P39639 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P100413 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P39963 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P121794 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 27220 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 27334 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55150 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33961914 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P39639 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P39639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30636432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30636432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10095150 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P39639 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P121794 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P39963 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P100413 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10095146 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10095147 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10095150 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10095148 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10095145 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909982 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10095147 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10095146 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909980 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909981 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909983 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909984 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909977 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909978 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16909979 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10095145 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P100413 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P39639 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33961914 - -
          Agilent Human 2 cDNA Microarray (G4101A) 588789 - -
          Agilent Human 2 cDNA Microarray (G4101A) 588789 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16909981 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095146 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095148 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16909983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16909979 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10095147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_058219_at - -
          Affymetrix Human Gene 1.0 ST Array 8002345 - -
          Affymetrix Human Gene 1.0 ST Array 8002346 - -
          Affymetrix Human Gene 1.1 ST Array 8002344 - -
          Affymetrix HT HG-U133+ PM Array Plate 231916_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1553947_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1558044_PM_s_at - -
          Affymetrix Human Genome U219 Array 11735184_at - -
          Affymetrix Human Genome U219 Array 11735184_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010751.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600183085.hg.1 ENST00000435634.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600183085.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600183085.hg.1 TC1600010751.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600183085.hg.1 NM_058219.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600183085.hg.1 ENST00000435634.2 Ensembl
          Human Genome U133 Plus 2.0 Array 1553947_at - -
          Human Genome U133 Plus 2.0 Array 231916_at - -
          Human Genome U133 Plus 2.0 Array 227696_PM_at - -
          Human Genome U133 Plus 2.0 Array 1558044_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 1553947_PM_at - -
          Human Genome U133 Plus 2.0 Array 227696_at - -
          Human Genome U133 Plus 2.0 Array 1558044_s_at - -
          Human Genome U133 Plus 2.0 Array 231916_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_058219_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231916_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558044_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227696_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553947_at1 - -
          Human Genome U95 Set 56286_at - -
          Human Genome U95 Set 56286_PM_at - -
          Human Genome U95 Set 63765_r_at - -
          Human Genome U95 Set 63764_i_at - -
          Human Genome U95 Set 63765_PM_r_at - -
          Human Genome U95 Set 63764_PM_i_at - -
          Human Genome U133 Set 227696_at - -
          Human Genome U133 Set 231916_PM_at - -
          Human Genome U133 Set 231916_at - -
          Human Genome U133 Set 227696_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053890 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053890 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 146084 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1670218 BC052252.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1670218 AK095542.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_32484988-S BC052252.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_32484988-S AK095542.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_32484988-S NM_058219.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_15247-S - -
          Illumina human-6 v1.0 expression beadchip 0003170497 - -
          Illumina humanRef-8 v1.0 expression beadchip 0003170497 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 70477 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_15247 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1670218 NM_058219.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1670218 BC052252.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1670218 AK095542.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1670218 NM_058219.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix