Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00991882_m1
          See other TMEM182 GE Assays ›
          Gene Symbol:
          TMEM182
          Gene Name:
          transmembrane protein 182
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 102736906 - 102843838 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00991882_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM182
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321343.1 NP_001308272.1 5 - 6 524 60
          NM_001321344.1 NP_001308273.1 4 - 5 397 60
          NM_001321345.1 NP_001308274.1 3 - 4 312 60
          NM_144632.4 NP_653233.4 3 - 4 523 60
          XM_006712287.1 XP_006712350.1 3 - 4 521 60
          XM_011510632.2 XP_011508934.1 3 - 4 521 60
          XM_017003376.1 XP_016858865.1 3 - 4 308 60
          GenBank mRNA AK054856.1 - 3 - 4 283 60
          AK129862.1 - 3 - 4 482 60
          AL832688.1 - 4 - 5 394 60
          AY358262.1 - 3 - 4 621 60
          BC073844.1 - 3 - 4 259 60
          BC107694.1 - 3 - 4 267 60
          BC130306.1 - 3 - 4 470 40146337 60
          BC130308.1 - 3 - 4 467 40146339 60
          CR936731.1 - 3 - 4 499 60
          CR936732.1 - 3 - 4 495 60
          CR936735.1 - 3 - 4 522 60
          CX163622.1 - 3 - 4 252 60
          DA355461.1 - 3 - 4 310 60
          DA908857.1 - 5 - 6 522 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200008682.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000629.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TMEM182

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 ENST00000409173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 ENST00000409528.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010020.hg.1 TC02000629.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 ENST00000409173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 ENST00000409528.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010021.hg.1 TC02000629.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 TC0200008682.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160726.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 TC0200008682.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200160725.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409173.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000409528.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000412401.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000454536.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000469971.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000484094.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000486293.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 ENST00000488134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq
          Affymetrix Human Clariom D Assay TC0200008682.hg.1 NM_144632.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0