Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00992268_m1
          See other TRIP12 GE Assays ›
          Gene Symbol:
          TRIP12
          Gene Name:
          thyroid hormone receptor interactor 12
          Gene Aliases:
          TRIP-12, ULF
          Chromosome Location:
          Chr.2: 229763837 - 229923234 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00992268_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIP12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001284214.1 NP_001271143.1 30 - 31 4639 104
          NM_001284215.1 NP_001271144.1 28 - 29 4464 104
          NM_001284216.1 NP_001271145.1 27 - 28 3685 104
          NM_004238.2 NP_004229.1 29 - 30 4495 104
          XM_005246954.4 XP_005247011.1 30 - 31 4747 104
          XM_005246955.4 XP_005247012.1 30 - 31 4746 104
          XM_005246956.4 XP_005247013.1 30 - 31 4723 104
          XM_005246957.4 XP_005247014.1 30 - 31 4698 104
          XM_005246958.4 XP_005247015.1 30 - 31 4666 104
          XM_005246960.4 XP_005247017.1 31 - 32 4660 104
          XM_005246961.3 XP_005247018.1 29 - 30 4597 104
          XM_005246962.4 XP_005247019.1 29 - 30 4594 104
          XM_006712852.3 XP_006712915.1 29 - 30 4550 104
          XM_011512180.2 XP_011510482.1 30 - 31 4700 104
          XM_011512181.2 XP_011510483.1 30 - 31 4698 104
          XM_011512183.2 XP_011510485.1 29 - 30 4554 104
          XM_017005272.1 XP_016860761.1 31 - 32 4806 104
          XM_017005273.1 XP_016860762.1 30 - 31 4673 104
          XM_017005274.1 XP_016860763.1 30 - 31 4621 104
          XM_017005275.1 XP_016860764.1 30 - 31 4676 104
          XM_017005276.1 XP_016860765.1 30 - 31 4681 104
          XM_017005277.1 XP_016860766.1 31 - 32 4821 104
          XM_017005278.1 XP_016860767.1 30 - 31 4636 104
          XM_017005279.1 XP_016860768.1 30 - 31 4693 104
          XM_017005280.1 XP_016860769.1 30 - 31 4682 104
          XM_017005281.1 XP_016860770.1 30 - 31 4616 104
          XM_017005282.1 XP_016860771.1 31 - 32 4659 104
          XM_017005283.1 XP_016860772.1 30 - 31 4648 104
          XM_017005284.1 XP_016860773.1 31 - 32 4642 104
          XM_017005285.1 XP_016860774.1 29 - 30 4584 104
          XM_017005286.1 XP_016860775.1 29 - 30 4616 104
          XM_017005287.1 XP_016860776.1 29 - 30 4582 104
          XM_017005288.1 XP_016860777.1 29 - 30 4544 104
          XM_017005289.1 XP_016860778.1 31 - 32 4579 104
          XM_017005290.1 XP_016860779.1 31 - 32 4576 104
          XM_017005291.1 XP_016860780.1 30 - 31 4510 104
          XM_017005292.1 XP_016860781.1 29 - 30 4498 104
          XM_017005293.1 XP_016860782.1 29 - 30 4513 104
          GenBank mRNA BC113891.1 - 27 - 28 3579 40083155 104
          BC114556.1 - 30 - 31 4533 40083165 104
          D28476.1 - 29 - 30 4425 104
          EU489742.1 - 28 - 29 4418 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015963.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002848.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIP12

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          DNA repair
          ubiquitin-dependent protein catabolic process
          cellular response to DNA damage stimulus
          embryo development
          negative regulation of histone H2A K63-linked ubiquitination
          negative regulation of double-strand break repair
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          thyroid hormone receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02045956.hg.1 ENST00000283943.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045956.hg.1 ENST00000389044.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045956.hg.1 ENST00000389045.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02045956.hg.1 NM_004238.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02045956.hg.1 TC02002848.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000283943.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000389044.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000389045.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284215.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284216.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_004238.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 TC0200015963.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000389044.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000283943.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 ENST00000389045.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284214.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_004238.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284215.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200217405.hg.1 NM_001284216.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0