Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00993074_m1
          See other SUPT20H GE Assays ›
          Gene Symbol:
          SUPT20H
          Gene Name:
          SPT20 homolog, SAGA complex component
          Gene Aliases:
          C13, C13orf19, FAM48A, FP757, P38IP, SPT20
          Chromosome Location:
          Chr.13: 37009312 - 37059714 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00993074_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SUPT20H
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014286.2 NP_001014308.2 14 - 15 1396 108
          NM_001278480.1 NP_001265409.1 14 - 15 1396 108
          NM_001278481.1 NP_001265410.1 14 - 15 1399 108
          NM_001278482.1 NP_001265411.1 14 - 15 1537 108
          NM_017569.3 NP_060039.1 14 - 15 1478 108
          XM_005266447.2 XP_005266504.1 14 - 15 1372 108
          XM_005266449.3 XP_005266506.1 14 - 15 1333 108
          XM_005266451.3 XP_005266508.1 14 - 15 1369 108
          XM_005266454.3 XP_005266511.1 13 - 14 1331 108
          XM_005266455.2 XP_005266512.1 14 - 15 1372 108
          XM_005266456.3 XP_005266513.1 13 - 14 1404 108
          XM_005266457.3 XP_005266514.1 13 - 14 1330 108
          XM_005266458.3 XP_005266515.1 13 - 14 1401 108
          XM_005266460.2 XP_005266517.1 14 - 15 1372 108
          XM_005266461.3 XP_005266518.1 14 - 15 1372 108
          XM_005266462.4 XP_005266519.1 14 - 15 1372 108
          XM_005266464.3 XP_005266521.1 14 - 15 1372 108
          XM_005266465.2 XP_005266522.1 14 - 15 1369 108
          XM_005266467.3 XP_005266524.1 14 - 15 1372 108
          XM_017020653.1 XP_016876142.1 14 - 15 1369 108
          XM_017020654.1 XP_016876143.1 13 - 14 1331 108
          XM_017020655.1 XP_016876144.1 13 - 14 1330 108
          XM_017020656.1 XP_016876145.1 13 - 14 1399 108
          XM_017020657.1 XP_016876146.1 14 - 15 1369 108
          XM_017020658.1 XP_016876147.1 13 - 14 1351 108
          XM_017020659.1 XP_016876148.1 13 - 14 1330 108
          XM_017020660.1 XP_016876149.1 14 - 15 1369 108
          XM_017020661.1 XP_016876150.1 13 - 14 1330 108
          GenBank mRNA AF093250.1 - 14 - 15 1344 108
          AJ130894.1 - 14 - 15 1434 108
          AK027554.1 - 13 - 14 1217 108
          AK225376.1 - 14 - 15 1332 108
          AK292376.1 - 14 - 15 1482 108
          AK304227.1 - 12 - 13 1263 108
          BC001145.1 - 15 - 16 1522 3533877 108
          BC030686.2 - 14 - 15 1371 4826077 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC13000571.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SUPT20H

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          gastrulation
          transcription coregulator activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000350612.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000356185.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000360252.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000464744.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000475892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000536874.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 ENST00000542180.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 NM_001014286.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 NM_017569.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13006532.hg.1 TC13000571.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0