Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004402.hg.1 |
ENST00000342972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004402.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004402.hg.1 |
TC0X000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004401.hg.1 |
ENST00000342972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004401.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004401.hg.1 |
TC0X000332.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00150743.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150743.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150743.hg.1 |
TC0X00007378.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00150743.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150743.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007378.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150744.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150744.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150744.hg.1 |
TC0X00007378.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00150744.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150744.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150745.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150745.hg.1 |
NM_014061.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150745.hg.1 |
TC0X00007378.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00150745.hg.1 |
ENST00000342972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150745.hg.1 |
NM_014061.4 |
Refseq |