Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00995846_g1
          See other PRKCI GE Assays ›
          Gene Symbol:
          PRKCI
          Gene Name:
          protein kinase C iota
          Gene Aliases:
          DXS1179E, PKCI, nPKC-iota
          Chromosome Location:
          Chr.3: 170222432 - 170305982 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00995846_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRKCI
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002740.5 NP_002731.4 13 - 14 1529 90
          XM_017006860.1 XP_016862349.1 12 - 13 1436 90
          GenBank mRNA AB451443.1 - 13 - 14 1264 90
          AK315342.1 - 13 - 14 1457 90
          BC022016.2 - 13 - 14 1524 4823853 90
          BC042405.1 - 1 - 2 295 90
          L18964.1 - 13 - 14 1528 90
          L33881.1 - 13 - 14 1468 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300009468.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000914.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRKCI

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein targeting to membrane
          cytoskeleton organization
          actin filament organization
          positive regulation of neuron projection development
          vesicle-mediated transport
          cell migration
          peptidyl-serine phosphorylation
          cellular response to insulin stimulus
          negative regulation of glial cell apoptotic process
          establishment of apical/basal cell polarity
          intracellular signal transduction
          eye photoreceptor cell development
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          establishment or maintenance of epithelial cell apical/basal polarity
          cell-cell junction organization
          positive regulation of glucose import
          secretion
          Golgi vesicle budding
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of glial cell proliferation
          membrane organization
          response to interleukin-1
          bicellular tight junction assembly
          positive regulation of protein localization to plasma membrane
          positive regulation of endothelial cell apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          protein binding
          ATP binding
          phospholipid binding
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03015967.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03015967.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03015967.hg.1 TC03000914.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 TC0300009468.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300168017.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000295797.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000476635.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000482353.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000483697.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000485837.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000488541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 ENST00000493761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq
          Affymetrix Human Clariom D Assay TC0300009468.hg.1 NM_002740.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0