Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00996208_s1
          See other MOP-1 GE Assays ›
          Gene Symbol:
          MOP-1
          Gene Name:
          MOP-1
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00996208_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MOP-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB014771.1 - 1 54 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CU456651.1 - 1 143 Chr. 4 - 81468836 - 81469398 [-] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RasGEF domain family member 1B RASGEF1B GPIG4 Sense Chr. 4 - 81426393 - 81471928 on NCBI Build GRCh38
          Downstream heterogeneous nuclear ribonucleoprotein D HNRNPD AUF1,
          AUF1A,
          HNRPD,
          P37,
          hnRNPD0
          884.0kb Sense Chr. 4 - 82353314 - 82373996 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P419028 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P419028 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 28677 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19384 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P419028 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P419028 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P419028 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_041845_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041846_at - -
          Affymetrix Human Gene 1.0 ST Array XR_041844_at - -
          Affymetrix Human Gene 1.0 ST Array 8101323 - -
          Affymetrix Human Gene 1.1 ST Array 8101322 - -
          Affymetrix HT HG-U133+ PM Array Plate 224277_PM_at - -
          Affymetrix Human Genome U219 Array 11761577_PM_at - -
          Affymetrix Human Genome U219 Array 11761577_at - -
          Human Genome U133 Plus 2.0 Array 224277_at - -
          Human Genome U133 Plus 2.0 Array 224277_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224277_at4 - -
          Human Genome U133 Set 224277_at - -
          Human Genome U133 Set 224277_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 10691547 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0