Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
ENST00000250377.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
ENST00000534898.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
ENST00000554896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
ENST00000556912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
NM_014672.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002387.hg.1 |
TC14002293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000250377.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000321130.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000534898.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000554896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000556121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000556711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
ENST00000556912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
NM_014672.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002388.hg.1 |
TC14002293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000250377.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000321130.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000534898.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000554896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000556121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
ENST00000556912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
NM_014672.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002389.hg.1 |
TC14002293.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000534898.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000604948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000605870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_001256679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_001256680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_014672.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
TC1400010595.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000534898.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000604948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
ENST00000605870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_001256679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_001256680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179123.hg.1 |
NM_014672.3 |
Refseq |