Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00996831_g1
          See other SLC18A2 GE Assays ›
          Gene Symbol:
          SLC18A2
          Gene Name:
          solute carrier family 18 member A2
          Gene Aliases:
          SVAT, SVMT, VAT2, VMAT2
          Chromosome Location:
          Chr.10: 117241073 - 117279430 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00996831_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC18A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003054.4 NP_003045.2 11 - 12 1235 112
          GenBank mRNA AK314997.1 - 11 - 12 1188 112
          BC030593.1 - 10 - 11 1493 112
          BC108927.1 - 10 - 11 1100 40025174 112
          BC108928.2 - 10 - 11 1100 40025175 112
          L09118.1 - 11 - 12 112
          L14269.1 - 11 - 12 1105 112
          L23205.1 - 11 - 12 1185 112
          X71354.1 - 11 - 12 1135 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009017.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000850.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC18A2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          serotonin transport
          chemical synaptic transmission
          neurotransmitter secretion
          aging
          locomotory behavior
          response to herbicide
          post-embryonic development
          response to zinc ion
          aminergic neurotransmitter loading into synaptic vesicle
          monoamine transport
          dopamine transport
          insulin secretion
          endocytic recycling
          sequestering of neurotransmitter
          response to cocaine
          glucose homeostasis
          response to starvation
          response to corticosterone
          negative regulation of neurotransmitter transport
          transmembrane transport
          cellular response to ammonium ion
          cellular response to xenobiotic stimulus
          neurotransmitter loading into synaptic vesicle
          negative regulation of reactive oxygen species biosynthetic process
          amine transmembrane transporter activity
          serotonin:sodium symporter activity
          monoamine transmembrane transporter activity
          enzyme binding
          heat shock protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010325.hg.1 ENST00000298472.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010325.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010325.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010325.hg.1 TC10000850.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 TC1000009017.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000161794.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
          Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0