Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00996919_m1
          See other PLA2G4A GE Assays ›
          Gene Symbol:
          PLA2G4A
          Gene Name:
          phospholipase A2 group IVA
          Gene Aliases:
          PLA2G4, cPLA2, cPLA2-alpha
          Chromosome Location:
          Chr.1: 186828900 - 186989896 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00996919_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLA2G4A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024420.2 NP_077734.1 6 - 7 627 122
          XM_005245267.3 XP_005245324.2 4 - 5 448 122
          XM_011509642.2 XP_011507944.1 6 - 7 578 122
          GenBank mRNA AK290336.1 - 6 - 7 578 122
          BC114340.2 - 6 - 7 524 40034995 122
          M68874.1 - 6 - 7 560 122
          M72393.1 - 6 - 7 547 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010939.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001607.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLA2G4A

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          luteolysis
          phospholipid metabolic process
          phosphatidic acid biosynthetic process
          platelet activating factor biosynthetic process
          icosanoid metabolic process
          aging
          positive regulation of cell population proliferation
          phospholipid catabolic process
          response to heat
          response to lithium ion
          response to organonitrogen compound
          arachidonic acid metabolic process
          positive regulation of bone mineralization
          positive regulation of vesicle fusion
          positive regulation of prostaglandin biosynthetic process
          positive regulation of fever generation
          response to lipopolysaccharide
          response to vitamin D
          cardiolipin acyl-chain remodeling
          phosphatidylglycerol acyl-chain remodeling
          phosphatidylinositol acyl-chain remodeling
          phosphatidylserine acyl-chain remodeling
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          response to hydrogen peroxide
          positive regulation of apoptotic process
          surfactant homeostasis
          icosanoid biosynthetic process
          decidualization
          arachidonic acid secretion
          response to glucocorticoid
          response to calcium ion
          response to methylmercury
          cellular response to antibiotic
          lysophospholipase activity
          phospholipase A2 activity
          calcium ion binding
          calcium-dependent phospholipid binding
          phospholipase A1 activity
          histone acetyltransferase binding
          calcium-dependent phospholipase A2 activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01024947.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01024947.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01024947.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01024947.hg.1 TC01001607.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 TC0100010939.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180465.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010939.hg.1 NM_024420.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0