Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000378438.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000486558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000535539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000535777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000538655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
ENST00000544975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002114.hg.1 |
TC0X000190.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
TC0X00007012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147689.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000378438.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000423649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000436783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000447485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000479120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000486558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
ENST00000487051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007012.hg.1 |
NM_005765.2 |
Refseq |