Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00999728_m1
          See other PICALM GE Assays ›
          Gene Symbol:
          PICALM
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          CALM, CLTH, LAP
          Chromosome Location:
          Chr.11: 85957171 - 86069881 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00999728_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PICALM
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001206946.1 NP_001193875.1 13 - 14 1708 148
          NM_007166.3 NP_009097.2 13 - 14 1729 148
          XM_005274322.2 XP_005274379.1 13 - 14 1713 148
          XM_005274323.2 XP_005274380.1 13 - 14 1713 148
          XM_005274324.2 XP_005274381.1 13 - 14 1713 148
          XM_005274325.2 XP_005274382.1 13 - 14 1713 148
          XM_005274326.2 XP_005274383.1 13 - 14 1713 148
          XM_005274327.2 XP_005274384.1 13 - 14 1713 148
          XM_005274328.2 XP_005274385.1 13 - 14 1713 148
          XM_005274329.3 XP_005274386.1 13 - 14 1713 148
          XM_005274331.2 XP_005274388.1 13 - 14 1713 148
          XM_005274334.2 XP_005274391.1 13 - 14 1713 148
          XM_005274337.2 XP_005274394.1 13 - 14 1713 148
          XM_006718699.2 XP_006718762.1 13 - 14 1692 148
          XM_006718700.2 XP_006718763.1 13 - 14 1692 148
          XM_006718701.3 XP_006718764.1 13 - 14 1692 148
          XM_017018381.1 XP_016873870.1 13 - 14 1692 148
          XM_017018385.1 XP_016873874.1 13 - 14 1713 148
          GenBank mRNA AB210017.1 - 13 - 14 1708 148
          AK291414.1 - 14 - 15 1596 148
          BC048259.1 - 13 - 14 1490 5537605 148
          U45976.1 - 13 - 14 1559 148

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PICALM

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          cell population proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          vesicle cargo loading
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          iron ion homeostasis
          protein-containing complex assembly
          clathrin-dependent endocytosis
          cell-cell adhesion
          iron ion import across plasma membrane
          positive regulation of neuron death
          positive regulation of amyloid-beta formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          protein binding
          1-phosphatidylinositol binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-7zrph:80/100.66.134.75:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix