Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001359_m1
          See other IL23R GE Assays ›
          Gene Symbol:
          IL23R
          Gene Name:
          interleukin 23 receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 67138638 - 67259979 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001359_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL23R
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144701.2 NP_653302.2 8 - 9 1133 90
          XM_005270516.2 XP_005270573.1 5 - 6 779 90
          XM_011540790.2 XP_011539092.1 8 - 9 1601 90
          XM_011540791.2 XP_011539093.1 8 - 9 1118 90
          GenBank mRNA AF461422.1 - 8 - 9 1133 90
          AM990313.1 - 5 - 6 819 90
          AM990314.1 - 4 - 5 449 90
          AM990315.1 - 4 - 5 464 90
          AM990316.1 - 4 - 5 453 90
          AM990319.1 - 3 - 4 303 90
          AM990320.1 - 3 - 4 307 90
          AM990322.1 - 2 - 3 146 90
          AM990323.1 - 5 - 6 605 90
          AM990324.1 - 5 - 6 539 90
          AM990325.1 - 5 - 6 610 90
          AM990326.1 - 4 - 5 227 90
          AM990333.1 - 3 - 4 302 90
          AM990337.1 - 5 - 6 539 90
          AY937250.1 - 6 - 7 940 90
          AY937251.1 - 5 - 6 779 90
          AY937252.1 - 5 - 6 779 90
          BC016829.1 - 3 - 4 336 90
          FM212241.1 - 6 - 7 769 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008648.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000739.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL23R

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          positive regulation of T-helper 1 type immune response
          inflammatory response
          positive regulation of activation of Janus kinase activity
          response to lipopolysaccharide
          negative regulation of interleukin-10 production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of natural killer cell proliferation
          response to interferon-gamma
          interleukin-23-mediated signaling pathway
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          regulation of tyrosine phosphorylation of STAT protein
          positive regulation of tyrosine phosphorylation of STAT protein
          positive regulation of memory T cell differentiation
          positive regulation of osteoclast differentiation
          defense response to Gram-negative bacterium
          positive regulation of NK T cell activation
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          interleukin-12 receptor binding
          protein binding
          interleukin-23 binding
          interleukin-23 receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000347310.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000395227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000431791.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000441823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000540775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 ENST00000540911.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 NM_144701.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01011908.hg.1 TC01000739.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000395227.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 ENST00000473881.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay TC0100008648.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 ENST00000425614.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 TC0100008648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 NM_144701.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 ENST00000347310.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100161546.hg.1 ENST00000425614.3 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0