Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001471_m1
          See other ITGB3 GE Assays ›
          Gene Symbol:
          ITGB3
          Gene Name:
          integrin subunit beta 3
          Gene Aliases:
          BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
          Chromosome Location:
          Chr.17: 47253842 - 47312711 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001471_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGB3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000212.2 NP_000203.2 2 - 3 181 82
          GenBank mRNA AY826979.1 - 2 - 3 170 82
          BC127666.1 - 2 - 3 181 40128462 82
          BC127667.1 - 2 - 3 185 40128464 82
          J02703.1 - 2 - 3 181 82
          KC120777.1 - 2 - 3 181 82
          KC120778.1 - 2 - 3 181 82
          M20311.1 - 2 - 3 177 82
          M35999.1 - 2 - 3 179 82
          U95204.1 - 2 - 3 161 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012274.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17002878.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITGB3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          platelet degranulation
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          blood coagulation
          positive regulation of endothelial cell migration
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of lipid storage
          smooth muscle cell migration
          cell growth
          cell migration
          platelet activation
          extracellular matrix organization
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          cell-substrate adhesion
          activation of protein kinase activity
          negative regulation of lipid transport
          heterotypic cell-cell adhesion
          substrate adhesion-dependent cell spreading
          tube development
          apolipoprotein A-I-mediated signaling pathway
          wound healing
          regulation of bone resorption
          viral entry into host cell
          vascular endothelial growth factor receptor signaling pathway
          mesodermal cell differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of lipoprotein metabolic process
          leukocyte migration
          negative chemotaxis
          angiogenesis involved in wound healing
          platelet aggregation
          virus receptor activity
          fibronectin binding
          protease binding
          protein disulfide isomerase activity
          platelet-derived growth factor receptor binding
          protein binding
          coreceptor activity
          fibroblast growth factor binding
          enzyme binding
          C-X3-C chemokine binding
          insulin-like growth factor I binding
          neuregulin binding
          identical protein binding
          vascular endothelial growth factor receptor 2 binding
          cell adhesion molecule binding
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17007416.hg.1 ENST00000262017.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007416.hg.1 ENST00000559488.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17007416.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007416.hg.1 TC17002878.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 ENST00000573377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay TC1700012274.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 ENST00000571680.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 TC1700012274.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 NM_000212.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 ENST00000559488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700202749.hg.1 ENST00000571680.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0