Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001715_g1
          See other KANSL3 GE Assays ›
          Gene Symbol:
          KANSL3
          Gene Name:
          KAT8 regulatory NSL complex subunit 3
          Gene Aliases:
          KIAA1310, NSL3, Rcd1
          Chromosome Location:
          Chr.2: 96593155 - 96638379 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001715_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KANSL3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001115016.2 NP_001108488.1 15 - 16 2085 126
          NR_047653.1 15 - 16 2146 126
          NR_047655.1 15 - 16 2072 126
          NR_047656.1 14 - 15 1994 126
          NR_047657.1 16 - 17 2163 126
          NR_047658.1 15 - 16 2062 126
          XM_005263982.2 XP_005264039.1 16 - 17 2162 126
          XM_005263984.1 XP_005264041.1 16 - 17 2162 126
          XM_011511454.2 XP_011509756.1 16 - 17 2162 126
          XM_011511455.2 XP_011509757.1 16 - 17 2131 126
          XM_011511456.2 XP_011509758.1 16 - 17 2165 126
          XM_011511457.2 XP_011509759.1 16 - 17 2084 126
          XM_011511459.2 XP_011509761.1 15 - 16 2087 126
          XM_011511460.2 XP_011509762.1 15 - 16 2084 126
          XM_011511462.2 XP_011509764.1 15 - 16 1991 126
          XM_011511463.2 XP_011509765.1 16 - 17 2175 126
          XM_011511464.2 XP_011509766.1 16 - 17 2234 126
          XM_011511466.2 XP_011509768.1 15 - 16 2097 126
          XM_011511467.2 XP_011509769.1 15 - 16 2017 126
          XM_017004471.1 XP_016859960.1 15 - 16 2069 126
          XM_017004473.1 XP_016859962.1 16 - 17 2175 126
          XM_017004474.1 XP_016859963.1 16 - 17 2070 126
          GenBank mRNA AB037731.1 - 16 - 17 2115 126
          AK302646.1 - 15 - 16 2013 126
          AK304011.1 - 14 - 15 1994 126
          AK314367.1 - 15 - 16 2022 126
          AY050169.1 - 15 - 16 2113 126
          BC024276.1 - 15 - 16 2022 126
          BC051763.1 - 14 - 15 1794 126
          BC063792.1 - 15 - 16 2015 6504387 126
          BC127820.1 - 3 - 4 462 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013531.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002110.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          histone H4-K5 acetylation
          histone H4-K8 acetylation
          histone H4-K16 acetylation
          histone acetyltransferase activity (H4-K5 specific)
          histone acetyltransferase activity (H4-K8 specific)
          histone acetyltransferase activity (H4-K16 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000354204.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000431828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000440133.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000444759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000447759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 ENST00000487070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 NM_001115016.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02034152.hg.1 TC02002110.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000354204.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000420155.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000431828.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000444759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000447759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000476534.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000487070.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000597150.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NM_001115016.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047653.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047654.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047655.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047656.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047657.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047658.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 TC0200013531.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000444759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000354204.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000420155.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000431828.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000447759.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000476534.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000487070.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 ENST00000597150.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NM_001115016.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047653.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047654.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047655.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047656.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047657.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200198990.hg.1 NR_047658.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0