Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01002433_m1
          See other BAZ2B GE Assays ›
          Gene Symbol:
          BAZ2B
          Gene Name:
          bromodomain adjacent to zinc finger domain 2B
          Gene Aliases:
          WALp4
          Chromosome Location:
          Chr.2: 159315312 - 159712439 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01002433_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BAZ2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289975.1 NP_001276904.1 32 - 33 6094 117
          NM_013450.3 NP_038478.2 33 - 34 6202 117
          XM_005246488.2 XP_005246545.2 34 - 35 6183 117
          XM_005246489.4 XP_005246546.2 33 - 34 7286 117
          XM_005246492.4 XP_005246549.2 33 - 34 7281 117
          XM_006712461.3 XP_006712524.2 37 - 38 6757 117
          XM_011511038.2 XP_011509340.1 34 - 35 6281 117
          XM_011511039.1 XP_011509341.1 31 - 32 5904 117
          XM_011511042.1 XP_011509344.1 32 - 33 5832 117
          XM_011511043.1 XP_011509345.1 30 - 31 5802 117
          XM_011511044.1 XP_011509346.1 31 - 32 5829 117
          XM_011511045.1 XP_011509347.1 31 - 32 5742 117
          XM_011511047.1 XP_011509349.1 33 - 34 6089 117
          XM_011511048.2 XP_011509350.1 33 - 34 6154 117
          XM_011511050.1 XP_011509352.1 31 - 32 5637 117
          XM_011511051.1 XP_011509353.1 31 - 32 5718 117
          XM_011511052.1 XP_011509354.1 30 - 31 5541 117
          XM_011511053.2 XP_011509355.1 32 - 33 5937 117
          XM_011511056.1 XP_011509358.1 22 - 23 3557 117
          XM_017003918.1 XP_016859407.1 37 - 38 6747 117
          XM_017003919.1 XP_016859408.1 37 - 38 6741 117
          XM_017003920.1 XP_016859409.1 39 - 40 6958 117
          XM_017003921.1 XP_016859410.1 34 - 35 6281 117
          XM_017003922.1 XP_016859411.1 34 - 35 6177 117
          XM_017003923.1 XP_016859412.1 32 - 33 5931 117
          XM_017003924.1 XP_016859413.1 31 - 32 5829 117
          XM_017003925.1 XP_016859414.1 33 - 34 6185 117
          XM_017003926.1 XP_016859415.1 33 - 34 6043 117
          XM_017003927.1 XP_016859416.1 33 - 34 6083 117
          XM_017003928.1 XP_016859417.1 33 - 34 5993 117
          XM_017003929.1 XP_016859418.1 33 - 34 5970 117
          XM_017003930.1 XP_016859419.1 33 - 34 5883 117
          XM_017003931.1 XP_016859420.1 33 - 34 5981 117
          XM_017003932.1 XP_016859421.1 30 - 31 5634 117
          XM_017003933.1 XP_016859422.1 32 - 33 5860 117
          XM_017003934.1 XP_016859423.1 32 - 33 5755 117
          XM_017003935.1 XP_016859424.1 31 - 32 5693 117
          XM_017003936.1 XP_016859425.1 31 - 32 5647 117
          XM_017003937.1 XP_016859426.1 26 - 27 4354 117
          GenBank mRNA AB032255.1 - 28 - 29 5575 117
          AB040909.2 - 32 - 33 6062 117
          AK074895.1 - 11 - 12 2320 117
          AL080173.1 - 4 - 5 642 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014719.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02005063.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BAZ2B

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 ENST00000343439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 ENST00000355831.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 ENST00000392782.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 ENST00000392783.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 NM_013450.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02038718.hg.1 TC02005063.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000392782.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000392783.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000474437.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000548440.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 NM_001289975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 NM_013450.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 TC0200014719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000474437.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000392782.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000392783.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 ENST00000548440.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 NM_001289975.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200207013.hg.1 NM_013450.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0