Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01003268_g1
          See other HPRT1 GE Assays ›
          Gene Symbol:
          HPRT1
          Gene Name:
          hypoxanthine phosphoribosyltransferase 1
          Gene Aliases:
          HGPRT, HPRT
          Chromosome Location:
          Chr.X: 134460145 - 134500668 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01003268_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HPRT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000194.2 NP_000185.1 3 - 4 492 148
          GenBank mRNA AK313435.1 - 3 - 4 472 148
          BC000578.2 - 3 - 4 364 3163726 148
          BT019350.1 - 3 - 4 325 148
          CR407645.1 - 3 - 4 325 148
          L29382.1 - 3 - 4 325 148
          L29383.1 - 3 - 4 325 148
          M31642.1 - 3 - 4 410 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00011326.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000636.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection GE Endogenous Controls Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HPRT1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          purine nucleotide biosynthetic process
          purine ribonucleoside salvage
          adenine salvage
          guanine salvage
          grooming behavior
          locomotory behavior
          cytolysis
          striatum development
          cerebral cortex neuron differentiation
          central nervous system neuron development
          GMP salvage
          IMP salvage
          dopamine metabolic process
          purine-containing compound salvage
          hypoxanthine salvage
          positive regulation of dopamine metabolic process
          GMP catabolic process
          IMP metabolic process
          hypoxanthine metabolic process
          lymphocyte proliferation
          dendrite morphogenesis
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          hypoxanthine phosphoribosyltransferase activity
          protein binding
          protein homodimerization activity
          guanine phosphoribosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008365.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008365.hg.1 ENST00000370796.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008365.hg.1 ENST00000462974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008365.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008365.hg.1 TC0X000636.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 TC0X00011326.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177948.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0