Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01003766_g1
          See other FBXW11 GE Assays ›
          Gene Symbol:
          FBXW11
          Gene Name:
          F-box and WD repeat domain containing 11
          Gene Aliases:
          BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
          Chromosome Location:
          Chr.5: 171861549 - 172006873 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01003766_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FBXW11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012300.2 NP_036432.2 3 - 4 743 81
          NM_033644.2 NP_387448.2 3 - 4 704 81
          NM_033645.2 NP_387449.2 2 - 3 641 81
          XM_005265855.4 XP_005265912.1 4 - 5 793 81
          XM_005265856.2 XP_005265913.1 5 - 6 763 81
          XM_005265857.2 XP_005265914.1 2 - 3 301 81
          XM_005265858.2 XP_005265915.1 4 - 5 805 81
          XM_017009279.1 XP_016864768.1 3 - 4 603 81
          XM_017009280.1 XP_016864769.1 4 - 5 619 81
          XM_017009281.1 XP_016864770.1 4 - 5 687 81
          GenBank mRNA AB014596.1 - 3 - 4 397 81
          AB033279.1 - 2 - 3 371 81
          AB033280.1 - 3 - 4 489 81
          AB033281.1 - 3 - 4 511 81
          AF176022.1 - 3 - 4 396 81
          AK294956.1 - 3 - 4 386 81
          AK314999.1 - 2 - 3 270 81
          BC026213.1 - 3 - 4 704 4419029 81

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FBXW11

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          protein dephosphorylation
          Wnt signaling pathway
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          protein destabilization
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          positive regulation of circadian rhythm
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          rhythmic process
          T cell receptor signaling pathway
          stress-activated MAPK cascade
          negative regulation of NIK/NF-kappaB signaling
          ubiquitin-protein transferase activity
          protein binding
          protein dimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline