Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01003822_m1
          See other FKBP1B GE Assays ›
          Gene Symbol:
          FKBP1B
          Gene Name:
          FK506 binding protein 1B
          Gene Aliases:
          FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase
          Chromosome Location:
          Chr.2: 24033205 - 24067743 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01003822_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FKBP1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054033.3 NP_473374.1 3 - 4 354 102
          NR_136536.1 5 - 6 511 102
          NR_136538.1 4 - 5 443 102
          NR_136539.1 3 - 4 241 102
          GenBank mRNA AF322070.1 - 3 - 4 246 102
          BC002614.1 - 3 - 4 297 3161896 102
          BM802489.1 - 3 - 4 354 102
          BU147063.1 - 3 - 4 192 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016413.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02004952.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FKBP1B

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          'de novo' protein folding
          smooth muscle contraction
          response to glucose
          negative regulation of heart rate
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          neuronal action potential propagation
          protein maturation by protein folding
          insulin secretion
          negative regulation of phosphoprotein phosphatase activity
          response to vitamin E
          ion transmembrane transport
          calcium-mediated signaling using intracellular calcium source
          protein refolding
          T cell proliferation
          response to hydrogen peroxide
          positive regulation of axon regeneration
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of sequestering of calcium ion
          regulation of cytosolic calcium ion concentration
          response to redox state
          regulation of ryanodine-sensitive calcium-release channel activity
          negative regulation of ryanodine-sensitive calcium-release channel activity
          chaperone-mediated protein folding
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          cell communication by electrical coupling involved in cardiac conduction
          regulation of cardiac conduction
          peptidyl-prolyl cis-trans isomerase activity
          signaling receptor binding
          ryanodine-sensitive calcium-release channel activity
          protein binding
          FK506 binding
          calcium channel inhibitor activity
          cyclic nucleotide binding
          transmembrane transporter binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02001454.hg.1 ENST00000380991.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02001454.hg.1 NM_054033.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02001454.hg.1 TC02004952.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 TC0200016413.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200221128.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380986.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000380991.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000421839.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000438630.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000452109.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 ENST00000496149.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_004116.4 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Affymetrix Human Clariom D Assay TC0200016413.hg.1 NM_054033.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 134740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix