Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01005834_m1
          See other ATRX GE Assays ›
          Gene Symbol:
          ATRX
          Gene Name:
          ATRX, chromatin remodeler
          Gene Aliases:
          ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
          Chromosome Location:
          Chr.X: 77504878 - 77786269 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01005834_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATRX
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138270.3 NP_612114.2 5 - 6 639 176
          XM_006724667.3 XP_006724730.1 4 - 5 492 176
          XM_017029604.1 XP_016885093.1 5 - 6 639 176
          XM_017029606.1 XP_016885095.1 5 - 6 740 176
          XM_017029609.1 XP_016885098.1 5 - 6 740 176
          XM_017029611.1 XP_016885100.1 4 - 5 492 176
          GenBank mRNA U72938.2 - 5 - 6 586 176

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00010126.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATRX

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA methylation
          DNA recombination
          nucleosome assembly
          DNA replication-independent nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatogenesis
          positive regulation of nuclear cell cycle DNA replication
          DNA damage response, signal transduction by p53 class mediator
          forebrain development
          replication fork processing
          positive regulation of telomere maintenance
          DNA duplex unwinding
          post-embryonic forelimb morphogenesis
          multicellular organism growth
          positive regulation of transcription by RNA polymerase II
          Sertoli cell development
          protein localization to chromosome, telomeric region
          seminiferous tubule development
          cellular response to hydroxyurea
          regulation of histone H3-K9 trimethylation
          negative regulation of telomeric RNA transcription from RNA pol II promoter
          positive regulation of telomeric RNA transcription from RNA pol II promoter
          negative regulation of maintenance of mitotic sister chromatid cohesion, telomeric
          DNA binding
          DNA helicase activity
          chromatin binding
          helicase activity
          protein binding
          ATP binding
          DNA translocase activity
          methylated histone binding
          histone binding
          metal ion binding
          chromo shadow domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000373344.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000395603.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000480283.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000623321.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624032.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624166.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624668.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000625063.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 NM_000489.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 NM_138270.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 TC0X00010126.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000623321.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000373344.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000395603.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000480283.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000625063.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624032.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624166.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 ENST00000624668.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 NM_000489.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169139.hg.1 NM_138270.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0