Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01006897_g1
          See other TXNIP GE Assays ›
          Gene Symbol:
          TXNIP
          Gene Name:
          thioredoxin interacting protein
          Gene Aliases:
          ARRDC6, EST01027, HHCPA78, THIF, VDUP1
          Chromosome Location:
          Chr.1: 145992426 - 145996631 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01006897_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TXNIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001313972.1 NP_001300901.1 3 - 4 566 72
          NM_006472.5 NP_006463.3 4 - 5 935 72
          XM_017000085.1 XP_016855574.1 4 - 5 884 72
          GenBank mRNA AK226090.1 - 4 - 5 847 72
          AK301148.1 - 4 - 5 457 72
          AK304670.1 - 3 - 4 566 72
          AY594328.2 - 4 - 5 570 72
          BC024305.1 - 4 - 5 464 72
          BC093702.1 - 4 - 5 644 7939547 72
          BC093704.1 - 4 - 5 644 7939549 72
          S73591.1 - 4 - 5 791 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015598.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001090.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TXNIP

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          protein import into nucleus
          cell cycle
          response to mechanical stimulus
          response to glucose
          keratinocyte differentiation
          response to estradiol
          response to progesterone
          regulation of cell proliferation
          response to drug
          response to hydrogen peroxide
          positive regulation of apoptotic process
          negative regulation of catalytic activity
          platelet-derived growth factor receptor signaling pathway
          response to calcium ion
          negative regulation of cell division
          cellular response to tumor cell
          enzyme inhibitor activity
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01017169.hg.1 TC01001090.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01017169.hg.1 ENST00000369317.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017169.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01017171.hg.1 TC01001090.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01017171.hg.1 ENST00000369317.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017171.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216683.hg.1 TC0100015598.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000486597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay TC0100015598.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000425134.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000488537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 ENST00000582401.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 NM_006472.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100216685.hg.1 TC0100015598.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0