Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01007074_g1
          See other ERBB2 GE Assays ›
          Gene Symbol:
          ERBB2
          Gene Name:
          erb-b2 receptor tyrosine kinase 2
          Gene Aliases:
          CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1
          Chromosome Location:
          Chr.17: 39688084 - 39728662 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01007074_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERBB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005862.2 NP_001005862.1 1 - 2 200 119
          NM_001289936.1 NP_001276865.1 1 - 2 200 119
          NM_001289938.1 NP_001276867.1 1 - 2 200 119
          GenBank mRNA AB025286.1 - 1 - 2 183 119
          AK295195.1 - 1 - 2 200 119
          BC110392.1 - 1 - 2 165 119
          BI755753.1 - 1 - 2 165 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC17000480.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK295195.1, NM_001005862.2, NM_001289936.1, NM_001289938.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ERBB2

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          protein phosphorylation
          signal transduction
          cell surface receptor signaling pathway
          enzyme linked receptor protein signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          peripheral nervous system development
          heart development
          neuromuscular junction development
          motor neuron axon guidance
          cell population proliferation
          phosphatidylinositol 3-kinase signaling
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          positive regulation of cell growth
          regulation of microtubule-based process
          negative regulation of immature T cell proliferation in thymus
          ERBB2 signaling pathway
          wound healing
          myelination
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          positive regulation of translation
          regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of transcription by RNA polymerase I
          positive regulation of transcription by RNA polymerase III
          protein autophosphorylation
          phosphatidylinositol phosphate biosynthetic process
          phosphatidylinositol-mediated signaling
          oligodendrocyte differentiation
          positive regulation of epithelial cell proliferation
          regulation of ERK1 and ERK2 cascade
          cellular response to growth factor stimulus
          cellular response to epidermal growth factor stimulus
          positive regulation of protein targeting to membrane
          regulation of cell motility
          RNA polymerase I core binding
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          transmembrane signaling receptor activity
          protein binding
          ATP binding
          protein C-terminus binding
          growth factor binding
          protein phosphatase binding
          identical protein binding
          ErbB-3 class receptor binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17005662.hg.1 ENST00000406381.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005662.hg.1 NM_001005862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005662.hg.1 TC17000480.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17005666.hg.1 ENST00000406381.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005666.hg.1 NM_001005862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005666.hg.1 TC17000480.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005664.hg.1 NM_001005862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005664.hg.1 TC17000480.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17005663.hg.1 ENST00000406381.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17005663.hg.1 NM_001005862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17005663.hg.1 TC17000480.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000428283.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000430460.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000438143.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015048.hg.1 ENST00000451851.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0