Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012744.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012744.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012744.hg.1 |
TC09000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166423.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166423.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166423.hg.1 |
TC0900009711.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900166423.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166423.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000304425.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
ENST00000448696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NM_176891.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009711.hg.1 |
NR_027054.1 |
Refseq |