Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01011176_g1
          See other RIPK3 GE Assays ›
          Gene Symbol:
          RIPK3
          Gene Name:
          receptor interacting serine/threonine kinase 3
          Gene Aliases:
          RIP3
          Chromosome Location:
          Chr.14: 24336021 - 24340036 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01011176_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RIPK3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006871.3 NP_006862.2 8 - 9 1485 71
          GenBank mRNA AF156884.1 - 8 - 9 1275 71
          AK075275.1 - 8 - 9 1440 71
          AY453693.1 - 8 - 9 1275 71
          AY494982.1 - 8 - 9 1268 71
          AY494983.1 - 8 - 9 1313 71
          BC062584.1 - 8 - 9 1416 5213027 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400010727.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14002317.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RIPK3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of T cell mediated cytotoxicity
          regulation of adaptive immune response
          cellular protein modification process
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          positive regulation of phosphatase activity
          activation of protein kinase activity
          regulation of interferon-gamma production
          T cell differentiation in thymus
          NIK/NF-kappaB signaling
          T cell homeostasis
          regulation of activated T cell proliferation
          protein autophosphorylation
          lymph node development
          spleen development
          thymus development
          positive regulation of NF-kappaB transcription factor activity
          protein homooligomerization
          protein heterooligomerization
          positive regulation of ligase activity
          positive regulation of oxidoreductase activity
          positive regulation of necroptotic process
          regulation of activation-induced cell death of T cells
          necroptotic process
          positive regulation of protein deacetylation
          apoptotic signaling pathway
          amyloid fibril formation
          positive regulation of reactive oxygen species metabolic process
          regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
          positive regulation of intrinsic apoptotic signaling pathway
          transcription coactivator activity
          protein kinase activity
          protein serine/threonine kinase activity
          NF-kappaB-inducing kinase activity
          protein binding
          ATP binding
          identical protein binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011180.hg.1 ENST00000216274.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011180.hg.1 NM_006871.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011180.hg.1 TC14002317.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 TC1400010727.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400181978.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000216274.9 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554338.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554569.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000554756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557253.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557624.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 ENST00000557662.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010727.hg.1 NM_006871.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0