Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01011327_g1
          See other DLL1 GE Assays ›
          Gene Symbol:
          DLL1
          Gene Name:
          delta like canonical Notch ligand 1
          Gene Aliases:
          DELTA1, DL1, Delta
          Chromosome Location:
          Chr.6: 170282200 - 170291075 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01011327_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DLL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005618.3 NP_005609.3 5 - 6 1198 84
          XM_005266934.3 XP_005266991.1 5 - 6 1664 84
          GenBank mRNA AF196571.1 - 5 - 6 1046 84
          AK314234.1 - 5 - 6 1070 84
          AY358892.1 - 5 - 6 765 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014031.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002342.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DLL1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          somitogenesis
          somite specification
          heart looping
          marginal zone B cell differentiation
          type B pancreatic cell development
          Notch signaling pathway
          Notch receptor processing
          determination of left/right symmetry
          compartment pattern specification
          regulation of blood pressure
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          proximal/distal pattern formation
          astrocyte development
          regulation of somitogenesis
          spinal cord development
          cerebellar molecular layer formation
          cerebellar Purkinje cell layer structural organization
          hemopoiesis
          cell differentiation
          regulation of cell adhesion
          negative regulation of epithelial cell differentiation
          negative regulation of interleukin-10 production
          negative regulation of glial cell apoptotic process
          organ growth
          regulation of growth
          negative regulation of cell differentiation
          negative regulation of epidermal cell differentiation
          negative regulation of inner ear auditory receptor cell differentiation
          negative regulation of myeloid cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of endocytosis
          positive regulation of transcription by RNA polymerase II
          lateral inhibition
          skeletal muscle tissue growth
          regulation of skeletal muscle tissue growth
          positive regulation of skeletal muscle tissue growth
          neuron fate specification
          inner ear development
          regulation of neurogenesis
          regulation of cell division
          retina development in camera-type eye
          retina morphogenesis in camera-type eye
          Notch signaling pathway involved in arterial endothelial cell fate commitment
          left/right axis specification
          nephron development
          proximal tubule development
          loop of Henle development
          clathrin-dependent endocytosis
          energy homeostasis
          endothelial tip cell fate specification
          neuronal stem cell population maintenance
          skin epidermis development
          regulation of vascular endothelial growth factor signaling pathway
          positive regulation of sprouting angiogenesis
          Notch binding
          calcium ion binding
          protein binding
          Tat protein binding
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06029348.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06029348.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06029348.hg.1 TC06002342.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06029349.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06029349.hg.1 NM_005618.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06029349.hg.1 TC06002342.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600196700.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196700.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600196700.hg.1 TC0600014031.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196700.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600196700.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196701.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196701.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600196701.hg.1 TC0600014031.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196701.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196701.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000366756.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 ENST00000630500.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014031.hg.1 NM_005618.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0