Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009654.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165915.hg.1 |
ENST00000156109.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165915.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165915.hg.1 |
TC0X00009654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00165915.hg.1 |
NM_015698.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165915.hg.1 |
ENST00000156109.6 |
Ensembl |