Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320143.1 | NP_001307072.1 | 7 - 8 | 1125 | 111 | |
NM_001320144.1 | NP_001307073.1 | 7 - 8 | 986 | 111 | ||
NM_025228.3 | NP_079504.2 | 7 - 8 | 1046 | 111 | ||
NR_109871.1 | 7 - 8 | 1046 | 111 | |||
XM_011510018.2 | XP_011508320.1 | 7 - 8 | 1030 | 111 | ||
XM_011510019.1 | XP_011508321.1 | 7 - 8 | 1030 | 111 | ||
XM_017002399.1 | XP_016857888.1 | 7 - 8 | 1030 | 111 | ||
XM_017002400.1 | XP_016857889.1 | 7 - 8 | 1030 | 111 | ||
GenBank mRNA | AK122792.1 | - | 7 - 8 | 1029 | 111 | |
AK125601.1 | - | 7 - 8 | 1125 | 111 | ||
AK225762.1 | - | 7 - 8 | 941 | 111 | ||
AK300555.1 | - | 8 - 9 | 891 | 111 | ||
AL049667.1 | - | 7 - 8 | 839 | 111 | ||
BC110302.1 | - | 7 - 8 | 1012 | 5219542 | 111 | |
BC130417.1 | - | 6 - 7 | 726 | 40146448 | 111 | |
BC144139.1 | - | 6 - 7 | 726 | 9052659 | 111 | |
BX648450.1 | - | 7 - 8 | 1025 | 111 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |