Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01012702_m1
          See other PDE3A GE Assays ›
          Gene Symbol:
          PDE3A
          Gene Name:
          phosphodiesterase 3A
          Gene Aliases:
          CGI-PDE, CGI-PDE A, CGI-PDE-A, HTNB
          Chromosome Location:
          Chr.12: 20369245 - 20684107 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01012702_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDE3A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000921.4 NP_000912.3 2 - 3 1048 75
          XM_017019420.1 XP_016874909.1 2 - 3 1029 75
          XM_017019421.1 XP_016874910.1 2 - 3 3746 75
          GenBank mRNA AJ005036.1 - 2 - 3 1030 75
          BC117369.1 - 2 - 3 1048 40125920 75
          BC117371.1 - 2 - 3 1048 40125922 75
          M91667.2 - 2 - 3 1037 75
          U36798.1 - 2 - 3 1008 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007037.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000227.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDE3A

          Gene Ontology Categories:

          Function(s) Process(es)

          oocyte maturation
          cAMP catabolic process
          lipid metabolic process
          response to xenobiotic stimulus
          diterpenoid metabolic process
          cAMP-mediated signaling
          cGMP-mediated signaling
          regulation of meiotic nuclear division
          negative regulation of apoptotic process
          negative regulation of vascular permeability
          positive regulation of vascular permeability
          response to cAMP
          positive regulation of oocyte development
          cellular response to cGMP
          cellular response to transforming growth factor beta stimulus
          3',5'-cyclic-AMP phosphodiesterase activity
          cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
          cAMP binding
          metal ion binding
          3',5'-cyclic-GMP phosphodiesterase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12002756.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12002756.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12002756.hg.1 NM_000921.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12002756.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12002756.hg.1 TC12000227.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000542675.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay TC1200007037.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 NM_001244683.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 TC1200007037.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 ENST00000359062.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 ENST00000544307.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 NM_000921.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200149223.hg.1 NM_001244683.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0