Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01013232_m1
          See other WNK1 GE Assays ›
          Gene Symbol:
          WNK1
          Gene Name:
          WNK lysine deficient protein kinase 1
          Gene Aliases:
          HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
          Chromosome Location:
          Chr.12: 752552 - 911452 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01013232_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WNK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001184985.1 NP_001171914.1 23 - 24 7006 132
          NM_014823.2 NP_055638.2 21 - 22 5482 132
          NM_018979.3 NP_061852.3 23 - 24 6226 132
          NM_213655.4 NP_998820.3 23 - 24 6982 132
          XM_006719003.2 XP_006719066.1 23 - 24 6548 132
          XM_011520997.2 XP_011519299.1 24 - 25 7836 132
          XM_011520998.2 XP_011519300.1 24 - 25 7787 132
          XM_011520999.2 XP_011519301.1 24 - 25 7787 132
          XM_011521000.2 XP_011519302.1 24 - 25 7790 132
          XM_011521001.2 XP_011519303.1 23 - 24 7511 132
          XM_011521002.2 XP_011519304.1 23 - 24 7328 132
          XM_011521003.2 XP_011519305.1 22 - 23 7052 132
          XM_011521004.2 XP_011519306.1 22 - 23 7049 132
          XM_011521005.2 XP_011519307.1 21 - 22 6274 132
          XM_011521006.2 XP_011519308.1 22 - 23 6467 132
          XM_011521007.2 XP_011519309.1 22 - 23 6464 132
          XM_011521008.2 XP_011519310.1 20 - 21 5729 132
          XM_011521009.2 XP_011519311.1 20 - 21 5726 132
          XM_017019834.1 XP_016875323.1 22 - 23 6089 132
          XM_017019835.1 XP_016875324.1 21 - 22 6008 132
          XM_017019836.1 XP_016875325.1 21 - 22 6005 132
          XM_017019837.1 XP_016875326.1 21 - 22 5813 132
          XM_017019838.1 XP_016875327.1 21 - 22 5810 132
          XM_017019839.1 XP_016875328.1 20 - 21 5729 132
          GenBank mRNA AB002342.2 - 21 - 22 4637 132
          AJ296290.1 - 23 - 24 5583 132
          BC141881.1 - 15 - 16 3050 132
          JQ358908.1 - 24 - 25 6822 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200006450.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000010.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WNK1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of systemic arterial blood pressure
          protein phosphorylation
          ion transport
          negative regulation of phosphatase activity
          peptidyl-threonine phosphorylation
          positive regulation of ion transmembrane transporter activity
          intracellular signal transduction
          protein autophosphorylation
          neuron development
          regulation of cellular process
          ion homeostasis
          negative regulation of protein serine/threonine kinase activity
          negative regulation of pancreatic juice secretion
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          chloride channel inhibitor activity
          potassium channel inhibitor activity
          protein kinase binding
          phosphatase binding
          protein serine/threonine kinase inhibitor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000252477.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000315939.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000340908.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000530271.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000535572.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000537501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 ENST00000537687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 NM_001184985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 NM_014823.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 NM_018979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 NM_213655.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000168.hg.1 TC12000010.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000315939.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000340908.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000530271.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000535572.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000537687.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_001184985.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_014823.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_018979.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_213655.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 TC1200006450.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000315939.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000340908.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000530271.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000535572.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 ENST00000537687.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_001184985.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_014823.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_018979.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200144280.hg.1 NM_213655.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline