Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01014815_g1
          See other IRF7 GE Assays ›
          Gene Symbol:
          IRF7
          Gene Name:
          interferon regulatory factor 7
          Gene Aliases:
          IMD39, IRF-7H, IRF7A, IRF7B, IRF7C, IRF7H
          Chromosome Location:
          Chr.11: 612555 - 615999 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01014815_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IRF7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001572.3 NP_001563.2 6 - 7 1046 94
          NM_004031.2 NP_004022.2 4 - 5 1125 94
          XM_005252906.3 XP_005252963.1 5 - 6 1111 94
          XM_005252907.3 XP_005252964.1 5 - 6 1108 94
          XM_011520066.2 XP_011518368.1 6 - 7 1022 94
          GenBank mRNA AF076494.1 - 4 - 5 1050 94
          BC136555.1 - 4 - 5 761 9020545 94
          U53830.1 - 5 - 6 968 94
          U73036.1 - 6 - 7 985 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009685.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001241.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IRF7

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          regulation of adaptive immune response
          transcription by RNA polymerase II
          cellular response to DNA damage stimulus
          response to virus
          immunoglobulin mediated immune response
          establishment of viral latency
          regulation of type I interferon production
          positive regulation of type I interferon production
          interferon-alpha production
          interferon-beta production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          regulation of MyD88-dependent toll-like receptor signaling pathway
          regulation of MyD88-independent toll-like receptor signaling pathway
          TRIF-dependent toll-like receptor signaling pathway
          MDA-5 signaling pathway
          innate immune response
          regulation of monocyte differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          positive regulation of type I interferon-mediated signaling pathway
          negative regulation of macrophage apoptotic process
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 ENST00000330243.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 ENST00000397562.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 ENST00000397566.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 ENST00000397574.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 ENST00000525445.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 NM_001572.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 NM_004031.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11015624.hg.1 TC11001241.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000330243.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000397566.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000397574.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000469048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000525445.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000525750.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000532788.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000533182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 NM_001572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 NM_004031.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 TC1100009685.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000330243.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000397566.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000397574.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000469048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000525445.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000525750.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000532788.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 ENST00000533182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 NM_001572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100172956.hg.1 NM_004031.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000330243.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000397566.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000397574.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000469048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000525445.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000525750.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000527160.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000532788.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000533182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 NM_001572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 NM_004031.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 TC1100009685.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000330243.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000397566.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000397574.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000469048.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000525445.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000525750.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000527160.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000532788.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 ENST00000533182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 NM_001572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100172957.hg.1 NM_004031.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline