Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01015582_g1
          See other TSNAXIP1 GE Assays ›
          Gene Symbol:
          TSNAXIP1
          Gene Name:
          translin associated factor X interacting protein 1
          Gene Aliases:
          TXI1
          Chromosome Location:
          Chr.16: 67806573 - 67828068 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01015582_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TSNAXIP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001288990.1 NP_001275919.1 12 - 13 1829 71
          NM_001288991.1 NP_001275920.1 10 - 11 1622 71
          NM_001288992.1 NP_001275921.1 8 - 9 1575 71
          NM_001288993.1 NP_001275922.1 9 - 10 1702 71
          NM_001288994.1 NP_001275923.1 7 - 8 1378 71
          NM_018430.3 NP_060900.2 12 - 13 2018 71
          XM_005256049.4 XP_005256106.1 12 - 13 2023 71
          XM_011523230.2 XP_011521532.1 11 - 12 2230 71
          XM_011523231.2 XP_011521533.1 11 - 12 2209 71
          XM_011523232.2 XP_011521534.1 12 - 13 2182 71
          XM_011523233.2 XP_011521535.1 12 - 13 2155 71
          XM_011523234.2 XP_011521536.1 11 - 12 2098 71
          XM_011523235.1 XP_011521537.1 11 - 12 1865 71
          XM_011523236.2 XP_011521538.1 9 - 10 1488 71
          XM_011523237.2 XP_011521539.1 9 - 10 1990 71
          XM_011523238.1 XP_011521540.1 8 - 9 1403 71
          XM_011523239.1 XP_011521541.1 9 - 10 1404 71
          XM_011523240.1 XP_011521542.1 10 - 11 1590 71
          XM_011523241.1 XP_011521543.1 9 - 10 1678 71
          XM_011523242.1 XP_011521544.1 8 - 9 1531 71
          XM_011523243.1 XP_011521545.1 8 - 9 1485 71
          XM_017023445.1 XP_016878934.1 9 - 10 1467 71
          XM_017023446.1 XP_016878935.1 9 - 10 1334 71
          XM_017023447.1 XP_016878936.1 9 - 10 1356 71
          XM_017023448.1 XP_016878937.1 9 - 10 1307 71
          XM_017023449.1 XP_016878938.1 10 - 11 1281 71
          XM_017023450.1 XP_016878939.1 11 - 12 1494 71
          XM_017023451.1 XP_016878940.1 11 - 12 1680 71
          XM_017023452.1 XP_016878941.1 8 - 9 1510 71
          XM_017023453.1 XP_016878942.1 10 - 11 1582 71
          XM_017023454.1 XP_016878943.1 9 - 10 1435 71
          XM_017023455.1 XP_016878944.1 9 - 10 1324 71
          GenBank mRNA AF132730.1 - 9 - 10 1506 71
          AK027245.1 - 12 - 13 1736 71
          AK301920.1 - 12 - 13 1602 71
          AK302300.1 - 1011 71
          BC111018.1 - 12 - 13 1772 5168384 71
          BU607826.1 - 4 - 5 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008173.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000546.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TSNAXIP1

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          biological_process
          cell differentiation
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16007828.hg.1 ENST00000388833.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16007828.hg.1 ENST00000415766.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16007828.hg.1 ENST00000431934.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16007828.hg.1 NM_018430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16007828.hg.1 TC16000546.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16007829.hg.1 ENST00000388833.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16007829.hg.1 ENST00000415766.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16007829.hg.1 NM_018430.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16007829.hg.1 TC16000546.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000388833.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000415766.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000431934.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000466164.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000487120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000561639.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000565339.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000568990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000569504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288990.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288991.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288992.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288993.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288994.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_018430.3 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 TC1600008173.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000466164.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000388833.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000415766.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000431934.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000487120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000561639.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000565339.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000568990.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288992.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 ENST00000569504.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288990.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288991.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288993.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_001288994.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600160975.hg.1 NM_018430.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0