Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016197.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016197.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016197.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016197.hg.1 |
TC08001253.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
ENST00000529918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010575.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000528666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
TC0800010575.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
NM_004056.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000317995.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000524872.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171836.hg.1 |
ENST00000528666.1 |
Ensembl |